Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503213

Sigma-Aldrich

Anti-Collagen XVIII α1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

COIA1, Collagen α-1(XVIII) chain, Endostatin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 178 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... COL18A1(80781)

General description

Endostatin is a naturally occurring 20kDa polypeptide derived from the C-terminal portion of type XVIII collagen. It functions as an anti-angiogenic cytokine that is expressed in various organs with the highest levels in liver, lung and kidney. The gene encoding it is localized on human chromosome 21q22.3. Anti-Collagen XVIII α1 Antibody detects endogenous levels of total Collagen XVIII α1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Collagen XVIII alpha1.

Immunogen Range: 801-850

Biochem/physiol Actions

Endostatin inhibits the proliferation and migration of endothelial cells. It blocks apoptosis of cells and G1 to S phase transition in the cell cycle. It inhibits angiogenesis by blocking the pro-angiogenic activities of VEGF and FGF-basic. Mutations in this gene lead to Knobloch Syndrome (KS), a rare autosomal recessive disorder.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S P Oh et al.
Genomics, 19(3), 494-499 (1994-02-01)
Types XV and XVIII collagen belong to a unique and novel subclass of the collagen superfamily for which we have proposed the name the MULTIPLEXIN family. Members of this class contain polypeptides with multiple triple-helical domains separated and flanked by
Extracellular matrix modulates angiogenesis in physiological and pathological conditions.
Neve A
BioMed Research International, 756078-756078 (2014)
Maria Rita Passos-Bueno et al.
Anais da Academia Brasileira de Ciencias, 78(1), 123-131 (2006-03-15)
Collagen XVIII, a proteoglycan, is a component of basement membranes (BMs). There are three distinct isoforms that differ only by their N-terminal, but with a specific pattern of tissue and developmental expression. Cleavage of its C-terminal produces endostatin, an inhibitor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service