Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502986

Sigma-Aldrich

Anti-C1QB, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Complement C1q subcomponent subunit B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... C1QB(713)

General description

Anti-C1QB Antibody detects endogenous levels of total C1QB protein.
The C1QB (complement C1q B chain) gene is mapped to human chromosome 1p36.12 and codes for the subcomponent of C1q protein, which is part of C1 complex of the classical pathway. The encoded protein is predominantly expressed in neurons and glia. It is localized to the growing synapses. The encoded protein is a hexameric molecule consisting of three distinct types of 18 polypeptide chains.

Immunogen

The antiserum was produced against synthesized peptide derived from human C1QB.

Immunogen Range: 161-210

Biochem/physiol Actions

The protein encoded by C1QB (complement C1q B chain) gene is involved in synapse formation and offers neuroprotection in the central nervous system. In response to a number of immune and nonimmune ligands, C1q protein activates proteases C1r and C1s in order to trigger classical complement pathway. C1q protein is regarded as a major component of the humoral innate immune defense. C1q carries out opsonophagocytosis, inflammation, and cytolysis to eliminate pathogenic microorganism and harmful apoptotic cells. Mutations in the gene causes systemic lupus erythematosus. Variation in the gene is also linked to rheumatoid schizophrenia, arthritis, amyloidotic polyneuropathy, metastasis of cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of C1QB gene polymorphism with schizophrenia in Armenian population
Zakharyan R
BMC Medical Genetics (2011)
Variation in complement protein C1q is not a major contributor to cognitive impairment in Parkinson?s disease
Sophia Carbutt
Neuroscience Letters, 594, 66?69-66?69 (2015)
Expression of recombinant human complement C1q allows identification of the C1r/C1s-binding sites
Isabelle Bally
Proceedings of the National Academy of Sciences of the USA, 110(21), 8650?8655-8650?8655 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service