Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502895

Sigma-Aldrich

Anti-MUNC-18A antibody produced in rabbit

affinity isolated antibody

Synonym(s):

N- Sec1, STB1, STXB1, STXBP1, Syntaxin binding protein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... STXBP1(6812)

General description

Anti-MUNC-18A Antibody detects endogenous levels of total MUNC-18A protein.
The gene STXBP1 (syntaxin binding protein 1) is mapped to human chromosome 9q34.1. The encoded protein is 594 amino acids long, which is expressed in all tissues with highest levels in retina and cerebellum.

Immunogen

The antiserum was produced against synthesized peptide derived from human MUNC-18a.

Immunogen Range: 279-328

Biochem/physiol Actions

The gene STXBP1 (syntaxin binding protein 1) encodes a regulatory protein of the components of the SNARE (soluble NSF/ N-ethylmaleimide-sensitive factor attachment protein receptor) complex that are crucial for neurotransmission. It is a neuronal protein that functions in synaptic vesicle release. Mutations in this gene have been associated with early infantile epileptic encephalopathy. It causes autosomal dominant mental retardation and nonsyndromic epilepsy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D A Swanson et al.
Genomics, 48(3), 373-376 (1998-04-18)
In a screen designed to identify genes expressed preferentially in retina, we identified a cDNA encoding the human ortholog of rat STXBP1 (n-Sec1, Munc-18-1, rbSec1), a protein implicated in vesicle trafficking and neurotransmitter release. This protein also has similarity to
Fadi F Hamdan et al.
Annals of neurology, 65(6), 748-753 (2009-06-27)
We sequenced genes coding for components of the SNARE complex (STX1A, VAMP2, SNAP25) and their regulatory proteins (STXBP1/Munc18-1, SYT1), which are essential for neurotransmission, in 95 patients with idiopathic mental retardation. We identified de novo mutations in STXBP1 (nonsense, p.R388X;
Hirotomo Saitsu et al.
Nature genetics, 40(6), 782-788 (2008-05-13)
Early infantile epileptic encephalopathy with suppression-burst (EIEE), also known as Ohtahara syndrome, is one of the most severe and earliest forms of epilepsy. Using array-based comparative genomic hybridization, we found a de novo 2.0-Mb microdeletion at 9q33.3-q34.11 in a girl

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service