Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502707

Sigma-Aldrich

Anti-RYR2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

RYR-2, cardiac muscle ryanodine receptor-calcium release channel, cardiac muscle-type ryanodine receptor, ryanodine receptor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Check Cart for Availability


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 564 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RYR2(6262)

General description

Anti-RYR2 Antibody detects endogenous levels of total RYR2 protein.
RYR2 (ryanodine receptor 2) is an intracellular Ca2+ release channel present on the sarcoplasmic reticulum (SR). It forms tetramer with four types 2 RyR polypeptides (RyR2) and four FK506 binding proteins (FKBP12.6).

Immunogen

The antiserum was produced against synthesized peptide derived from human RyR2.

Immunogen Range: 2774-2823

Application

Anti-RYR2 antibody produced in rabbit is suitable for immunohistochemistry and indirect ELISA.

Biochem/physiol Actions

RYR2 (ryanodine receptor 2) provides guidance to the release and transport of Ca2+ from sarcoplasmic reticulum (SR) to the cytoplasm during cardiac muscle excitation-contraction (EC) coupling. Protein kinase A (PKA) phosphorylated RYR2 separates (FKBP12.6) to regulate the channel open probability (Po). During the process, a small portion of Ca2+ enter into the cell through the L-type Ca2+ channel, which further activates the RyR2 channel upon membrane depolarization. The activated RyR2 channel releases a large amount of Ca2+ from the SR and subsequent muscle contraction. Missense mutations in this gene cause Kazakh idiopathic ventricular tachycardia and arrhythmogenic right ventricular dysplasia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dawei Jiang et al.
Proceedings of the National Academy of Sciences of the United States of America, 101(35), 13062-13067 (2004-08-24)
The cardiac ryanodine receptor (RyR2) governs the release of Ca2+ from the sarcoplasmic reticulum, which initiates muscle contraction. Mutations in RyR2 have been linked to ventricular tachycardia (VT) and sudden death, but the precise molecular mechanism is unclear. It is
Ainur Akilzhanova et al.
PloS one, 9(6), e101059-e101059 (2014-07-01)
Channelopathies, caused by disturbed potassium or calcium ion management in cardiac myocytes are a major cause of heart failure and sudden cardiac death worldwide. The human ryanodine receptor 2 (RYR2) is one of the key players tightly regulating calcium efflux
Nathalie Roux-Buisson et al.
Heart rhythm, 11(11), 1999-2009 (2014-07-22)
Arrhythmogenic right ventricular cardiomyopathy/dysplasia (ARVC/D) is a genetic disease predominantly caused by desmosomal gene mutations that account for only ~50% of cases. Ryanodine receptor 2 (RYR2) gene mutations usually cause catecholaminergic polymorphic ventricular tachycardia but have been associated with a
S O Marx et al.
Cell, 101(4), 365-376 (2000-06-01)
The ryanodine receptor (RyR)/calcium release channel on the sarcoplasmic reticulum (SR) is the major source of calcium (Ca2+) required for cardiac muscle excitation-contraction (EC) coupling. The channel is a tetramer comprised of four type 2 RyR polypeptides (RyR2) and four
Xiaojing Luo et al.
Frontiers in cell and developmental biology, 8, 772-772 (2020-09-10)
In adult cardiomyocytes (CMs), the type 2 ryanodine receptor (RYR2) is an indispensable Ca2+ release channel that ensures the integrity of excitation-contraction coupling, which is fundamental for every heartbeat. However, the role and importance of RYR2 during human embryonic cardiac

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service