Skip to Content
Merck
All Photos(3)

Documents

SAB4502616

Sigma-Aldrich

Anti-NF-κB p65 (Acetyl-Lys310) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NFKB3, RELA, TF65, nuclear factor NF-κ-B p65 subunit, p65

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

acetylation (Lys310)

Gene Information

human ... RELA(5970)

General description

Anti-NF-κB p65 (Acetyl-Lys310) Antibody detects endogenous levels of total NF-κB p65 (Acetyl-Lys310) protein.
RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A) codes for the important constituent of the NF-κB (nuclear factor κ-B) complex. It is located on human chromosome 11q13.1. It is also called as nuclear factor κB subunit 3 (NFKB3).

Immunogen

The antiserum was produced against synthesized peptide derived from human NF-kappaB p65 around the acetylated site of Lys310.

Immunogen Range: 275-324

Application

Anti-NF-κB p65 (Acetyl-Lys310) antibody has been used in western blotting.

Biochem/physiol Actions

RELA (v-rel avian reticuloendotheliosis viral oncogene homolog A) participates in nuclear factor κ-B (NFKB) signaling pathway. It also controls inflammation. Alterations in the gene results in schizophrenia and startle reactions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

17β-Estradiol Attenuates Hematoma Expansion through ERα/Sirt1/NF-?B Pathway in Hyperglycemic Intracerebral Hemorrhage Mice
Zheng Y, et al.
Stroke, 46(2), 485-485 (2015)
Variants of the RELA gene are associated with schizophrenia and their startle responses.
Hashimoto R, et al.
Neuropsychopharmacology, 36(9), 1921-1921 (2011)
Genome-wide association analysis of soluble ICAM-1 concentration reveals novel associations at the NFKBIK, PNPLA3, RELA, and SH2B3 loci.
Pare G, et al.
PLoS Genetics, 7(4), e1001374-e1001374 (2011)
Yun Zheng et al.
Stroke, 46(2), 485-491 (2014-12-20)
17β-estradiol (E2) has been reported to reduce bleeding and brain injury in experimental intracerebral hemorrhage (ICH) model. However, it is not clear if E2 can prevent early hematoma expansion (HE) induced by hyperglycemia in acute ICH. The aim of this
Chun-Cheih Chao et al.
Cell, 179(7), 1483-1498 (2019-12-10)
Metabolism has been shown to control peripheral immunity, but little is known about its role in central nervous system (CNS) inflammation. Through a combination of proteomic, metabolomic, transcriptomic, and perturbation studies, we found that sphingolipid metabolism in astrocytes triggers the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service