Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502562

Sigma-Aldrich

Anti-RAD21 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Double-strand-break repair protein rad21 homolog, Hhr21, NXP-1, Nuclear matrix protein 1, SCC1 homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 71 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAD21(5885)

General description

Anti-RAD21 Antibody detects endogenous levels of total RAD21 protein.
RAD21 (RAD21 homolog, S. pombe) is a novel nuclear protein located on chromosome 8q24.

Immunogen

The antiserum was produced against synthesized peptide derived from human RAD21.

Immunogen Range: 521-570

Application

Anti-RAD21, C-Terminal antibody produced in rabbit is suitable for indirect ELISA and western blot applications.

Biochem/physiol Actions

RAD21 (RAD21 homolog, S. pombe) plays a crucial role in normal cell division. It acts as a main component of the cohesin complex for the proper chromosomal arrangement during cell division. The protein possesses two mitotic cleavage sites for the attachment of separase. It has been shown experimentally that at the beginning of anaphase stage, a caspase-like Esp1/separase attaches to the cleavage site of RAD21 and initiate sister chromatid separation, which is an essential step for the completion of cytokinesis. During double strand DNA damage repair, it helps the two DNA strands to adhere with each other. Mutation in RAD21 causes a congenital phenotype, cohesinopathy in humans characterized with growth retardation and minor skeletal anomalies.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kati P Porkka et al.
Genes, chromosomes & cancer, 39(1), 1-10 (2003-11-07)
To detect genes that are overexpressed in prostate cancer, a subtracted cDNA library was first constructed from the PC-3 cell line and subsequently screened by using cDNA microarray hybridization. Sixty-eight genes were found to be overexpressed (ratio>3) in PC-3. Half
Feng Chen et al.
The Journal of biological chemistry, 277(19), 16775-16781 (2002-03-05)
Caspases are a conserved family of proteases that play a critical role in the execution of apoptosis by cleaving key cellular proteins at Asp residues and modifying their function. Using an expression cloning strategy we recently developed, we isolated human
Matthew A Deardorff et al.
American journal of human genetics, 90(6), 1014-1027 (2012-05-29)
The evolutionarily conserved cohesin complex was originally described for its role in regulating sister-chromatid cohesion during mitosis and meiosis. Cohesin and its regulatory proteins have been implicated in several human developmental disorders, including Cornelia de Lange (CdLS) and Roberts syndromes.
Debananda Pati et al.
Molecular and cellular biology, 22(23), 8267-8277 (2002-11-06)
Rad21 is one of the major cohesin subunits that holds sister chromatids together until anaphase, when proteolytic cleavage by separase, a caspase-like enzyme, allows chromosomal separation. We show that cleavage of human Rad21 (hRad21) also occurs during apoptosis induced by

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service