Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502553

Sigma-Aldrich

Anti-Paxillin antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PAX-1, PAXI, Paired box protein Pax-1, paxillin, paxillin α

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 64 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PXN(5829)

General description

Anti-Paxillin Antibody detects endogenous levels of total Paxillin protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Paxillin.

Immunogen Range: 85-134

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Sivaraman et al.
Cell biology international, 46(3), 391-402 (2021-12-10)
The surface properties of three-dimensional scaffolds are improved by coating or covalently linking certain adhesion-promoting proteins or peptides. In the present study, the effect of type I collagen-derived peptide (GKNGDDGEA) on adhesion and proliferation of HaCaT keratinocytes and NIH3T3 murine
K Sivaraman et al.
Life sciences, 226, 140-148 (2019-04-16)
The surface modification of biomaterials with matrikines for tissue engineering application is one of the recent approaches to improve their biocompatibility. In an earlier study, a peptide containing 21 amino acid isolated from bovine tendon collagen was shown to promote
Katarzyna A Makowska et al.
Cell reports, 13(10), 2118-2125 (2015-12-17)
We investigated the myosin expression profile in prostate cancer cell lines and found that Myo1b, Myo9b, Myo10, and Myo18a were expressed at higher levels in cells with high metastatic potential. Moreover, Myo1b and Myo10 were expressed at higher levels in
Yashavantha L Vishweshwaraiah et al.
Nature communications, 12(1), 6615-6615 (2021-11-18)
Advances in protein design have brought us within reach of developing a nanoscale programming language, in which molecules serve as operands and their conformational states function as logic gates with precise input and output behaviors. Combining these nanoscale computing agents
Andrey V Cybulsky et al.
Physiological reports, 12(1), e15897-e15897 (2024-01-02)
SLK controls the cytoskeleton, cell adhesion, and migration. Podocyte-specific deletion of SLK in mice leads to podocyte injury as mice age and exacerbates injury in experimental focal segment glomerulosclerosis (FSGS; adriamycin nephrosis). We hypothesized that adhesion proteins may be substrates

Articles

High titer lentiviral particles including beta-actin, alpha-tubulin and vimentin used for live cell analysis of cytoskeleton structure proteins.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service