Skip to Content
Merck
All Photos(2)

Key Documents

SAB4502492

Sigma-Aldrich

Anti-NDRG3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Protein NDRG3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€347.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 41 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NDRG3(57446)

General description

NDRG3 (N-myc downstream-regulated gene 3 protein) is a novel member of N-myc down-regulated gene (NDRG) family mapped to human chromosomal location 20q11.21-q11.23. It is abundantly expressed in brain, prostate, ovary, and testis tissues. It is localized to the outer layers of seminiferous epithelium.

Immunogen

The antiserum was produced against synthesized peptide derived from human NDRG3.

Immunogen Range: 206-255

Application

Anti-NDRG3 antibody produced in rabbit is suitable for immunohistochemistry and western blot.

Biochem/physiol Actions

N-myc downstream-regulated gene 3 protein (NDRG3) is involved in prostate cancer cell growth in vitro and in vivo. It acts as an androgen regulated and prostate enriched gene during controlling of cancer cell growth. Thus, overexpression of NDRG3 has been linked to the malignant phenotype of tumors. In hepatitis B virus (HBV)-related hepatocellular carcinoma cell line, NDRG3 gene is inversely expressed with the miR-122 expression. Thus, it has been reported that in conjugation with miR-122, NDRG3 can be a potential therapeutic marker for hepatocellular carcinoma (HCC). Prominent localization of NDRG3 in the epithelium cells indicate its role in spermatogenesis. It also plays an important role in lactate-induced hypoxia signaling pathway in hypoxia. Its protein level expression in cell is negatively regulated by oxygen, whereas, whereas excess lactate produced under prolonged hypoxia blocks its proteasomal degradation by binding to NDRG3.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kyung Chan Park et al.
BMB reports, 48(6), 301-302 (2015-05-06)
Hypoxia is associated with many pathological conditions as well as the normal physiology of metazoans. We identified a lactate-dependent signaling pathway in hypoxia, mediated by the oxygen- and lactate-regulated protein NDRG family member 3 (NDRG3). Oxygen negatively regulates NDRG3 expression
Xianghu Qu et al.
Molecular and cellular biochemistry, 229(1-2), 35-44 (2002-04-09)
NDRG1 (N-Myc downstream regulated) is upregulated during cell differentiation, repressed by N-myc and c-myc in embryonic cells, and suppressed in several tumor cells. A nonsense mutation in the NDRG1 gene has been reported to be causative for hereditary motor and
W Zhao et al.
Biochimica et biophysica acta, 1519(1-2), 134-138 (2001-06-19)
We report the cloning and expression pattern of a novel N-myc downstream-regulated gene 3 (NDRG3), located on human chromosome 20q11.21-11.23. The NDRG3 cDNA is 2588 base pair in length, encoding a 363 amino acid polypeptide highly related to mouse Ndr3
Weiqun Wang et al.
International journal of cancer, 124(3), 521-530 (2008-11-01)
NDRG3 belongs to the N-myc down-regulated gene (NDRG) family that contains 4 paralogs: NDRG1, -2, -3 and -4. The function of NDRG3 and its relationship to cancer has not been studied. We herein report our examination of the expression and
Chun-Guang Fan et al.
Oncology reports, 26(5), 1281-1286 (2011-07-05)
microRNAs (miRNAs) are short, non-coding RNAs with post-transcriptional regulatory functions that participate in diverse biological pathways. miR-122, a liver-specific miRNA, has been found to be down-regulated in hepatocellular carcinoma (HCC) and HCC-derived cell lines. In this study, miR-122 was down-regulated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service