Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502371

Sigma-Aldrich

Anti-PKD1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KPCD1, PKC-mu, PKCM, PKD, PRKCM

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 101 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... KPCD1(5587)

Related Categories

General description

Anti-PKD1 Antibody detects endogenous levels of total PKD1 protein.
Polycystic kidney disease 1/polycystin-1 (PKD1/PC1) gene comprises of 46 exons and spans approximately 52 kb of genomic sequence. Polycystin-1 (PC1) is expressed at high levels in the brain and moderately in the kidney in adult tissue. It is present in the primary cilium of most mammalian cells. PC1 is also located at focal adhesions, desmosomes, and adherens junctions in the plasma membrane. Anti-PKD1 antibody detects endogenous levels of total PKD1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human PKD1/PKC mu.

Immunogen Range: 171-220

Application

Anti-PKD1 antibody produced in rabbit has been used in western blot analysis(1:500).[1]

Biochem/physiol Actions

Polycystic kidney disease 1/polycystin-1 (PKD1/PC1) plays a regulatory role in cell migration and cell proliferation through mammalian targets of rapamycin (mTOR) and Janus kinase (JAK) signals. PKD1 inhibits cell migration and in some situations, acts as a tumor suppressor protein in the lung cancer cell line (A549). Before birth, polycystin-1 is the most active protein in kidney cells, and it interacts and works with polycystin-2 (PKD2). Mutations in the PKD1 gene are responsible for 85 percent of polycystic kidney diseases (PKDs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PKD1 (polycystic kidney disease 1 (autosomal dominant))
Tan YC and Rennert H
Atlas of Genetics and Cytogenetics in Oncology and Haematology (2012)
Se-Hwa Jung et al.
International journal of molecular sciences, 22(9) (2021-06-03)
Polycystic Kidney Disease (PKD) is a disorder that affects the kidneys and other organs, and its major forms are encoded by polycystin-1 (PC1) and polycystin-2 (PC2), as PKD1 and PKD2. It is located sandwiched inside and outside cell membranes and
Giulio Innamorati et al.
Cell death & disease, 8(7), e2930-e2930 (2017-07-14)
Chorionic stem cells represent a promising opportunity for regenerative medicine. A deeper understanding of the stimuli that regulate their physiology, could lead to innovative clinical approaches. We revealed the presence of multiple sphingosine-1-phosphate (S1P) receptor isoforms in chorion-derived mesenchymal stem
Yu Zhang et al.
Journal of cellular physiology, 234(10), 18513-18523 (2019-03-27)
Cytoskeleton which includes microtubule and actin filaments plays important roles during mammalian oocyte maturation. In the present study, we showed that protein kinase C mu (PKC mu) was one potential key molecule which affected cytoskeleton dynamics in mouse oocytes. Our
Yu Zhang et al.
Aging, 10(12), 3736-3744 (2018-12-18)
Protein kinase D (PKD) subfamily which includes PKD1, PKD2 and PKD3 is a novel family of serine/threonine kinases. PKD has been widely implicated in the regulation of multiple physiological effects including immune responses, apoptosis and cell proliferation. However, the roles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service