Skip to Content
Merck
All Photos(3)

Key Documents

SAB4502062

Sigma-Aldrich

Anti-PAK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

α-PAK, PAK-1, Serine/threonine-protein kinase PAK 1, p21-activated kinase 1, p65-PAK

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 60 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PAK1(5058)

General description

Anti-PAK1 antibody detects endogenous levels of total PAK1 protein.
The PAK1 (p21 (RAC1) activated kinase 1) gene is mapped to human chromosome 11q13.5-q14.1. It belongs to the family of serine/threonine kinases and the gene is evolutionarily conserved.

Immunogen

The antiserum was produced against synthesized peptide derived from human PAK1.

Immunogen Range: 178-227

Biochem/physiol Actions

PAK1 (p21 (RAC1) activated kinase 1) maintains the cytoskeleton and cellular morphology. It is associated with cell cycle, cell motility, survival, and cell death. PAK1 is upregulated in different types of cancer. PAK1 is found to be involved in growth, invasion, and metastasis of cancer cells, as it affects the action of pro-apoptotic signals. In neurodegenerative diseases, such as Huntington′s disease and Alzheimer′s disease, and mental retardation, abnormal regulation of PAK1 is observed. PAK1 induces MAPK (mitogen activated protein kinase) signal pathway and cytoskeletal remodeling in breast cancer, thereby serving as an oncogene. PAK1 is known to significantly participate in the development, maturation and plasticity of the nervous system.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Threonine 209 phosphorylation on RUNX3 by Pak1 is a molecular switch for its dualistic functions.
Kumar A
Oncogene, 35(37), 4857-4865 (2016)
Expression of p21-activated kinases 1 and 3 is altered in the brain of subjects with depression.
Fuchsova B
Neuroscience, 333, 331-344 (2016)
Down-regulation of p21-activated serine/threonine kinase 1 is involved in loss of mesencephalic dopamine neurons.
Kim H
Molecular Brain, 9(1):45, 1-13 (2016)
Combined array comparative genomic hybridization and tissue microarray analysis suggest PAK1 at 11q13.5-q14 as a critical oncogene target in ovarian carcinoma.
Schraml P
The American Journal of Pathology, 163(3), 985-992 (2003)
MicroRNA-494 inhibits breast cancer progression by directly targeting PAK1.
Zhan MN
Cell Death & Disease, 8(1), 1-11 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service