Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501470

Sigma-Aldrich

Anti-5-HT-1B antibody produced in rabbit

affinity isolated antibody

Synonym(s):

5-HT-1B, 5-HT-1D-β, 5-HT1B, 5-hydroxytryptamine receptor 1B, Serotonin receptor 1B

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Check Cart for Availability


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 43 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HTR1B(3351)

Related Categories

General description

5-HT-1B/HTR1B (5-hydroxytryptamine receptor 1B) is a presynaptic and postsynaptic receptor, present in the basal ganglia, hippocampus and cortex. It is located on human chromosome 6q14.1.
Anti-5-HT-1B Antibody detects endogenous levels of total 5-HT-1B protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human 5-HT-1B.

Immunogen Range: 201-250

Biochem/physiol Actions

5-HT-1B/HTR1B (5-hydroxytryptamine receptor 1B) plays an important role in locomotion, feeding and thermoregulation and also in arterial contractile regulation mechanisms. It participates in regulating the effects of serotonin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polymorphisms in the 5′-untranslated region of the human serotonin receptor 1B (HTR1B) gene affect gene expression.
Duan J, et al.
Molecular Psychiatry, 8(11), 901-910 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service