Skip to Content
Merck
All Photos(1)

Key Documents

SAB4501427

Sigma-Aldrich

Anti-HOXA6 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HOX1B, Homeobox protein Hox-A6, Hox-1B

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 26 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HXA6(3203)

General description

Anti-HOXA6 Antibody detects endogenous levels of total HOXA6 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human HOXA6.

Immunogen Range: 101-150

Application

Anti-HOXA6 antibody produced in rabbit is suitable for indirect ELISA and western blot.

Biochem/physiol Actions

The members of HOX (Homeobox) gene family clusters with each other to form a complex. In HOXA6 (homeobox A6), there are eight homeoboxes of 90kb DNA in the cluster. It is expressed in the human adult small intestinal mucosa. It acts as transcription factors in defining gradients of cellular differentiation. It mainly functions in hemopoietic progenitor cell development, embryonic and fetal development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D Acampora et al.
Nucleic acids research, 17(24), 10385-10402 (1989-12-25)
We report the identification of 10 new human homeobox sequences. Altogether, we have isolated and sequenced 30 human homeoboxes clustered in 4 chromosomal regions called HOX loci. HOX1 includes 8 homeoboxes in 90 kb of DNA on chromosome 7. HOX2
E Boncinelli et al.
Genome, 31(2), 745-756 (1989-01-01)
We report the genomic organization of 20 human class I homeoboxes and the predicted primary sequence of the encoded homeodomains. These homeoboxes are clustered in four complex HOX loci on chromosomes 2, 7, 12, and 17. The homeoboxes of one
D Duboule et al.
The EMBO journal, 8(5), 1497-1505 (1989-05-01)
This paper reports the cloning of the fourth major murine homeogene complex, HOX-5. The partial characterization of this gene cluster revealed the presence of two novel genes (Hox-5.2, Hox-5.3) located at the 5' extremity of this complex. In situ hybridization
J R Walters et al.
Gastroenterology, 113(2), 472-477 (1997-08-01)
Different digestive enzymes and transporters are present in the duodenum, jejunum, and ileum, but the factors determining region-specific gene expression are not yet understood. Homeobox transcription factors are important in defining gradients of cellular differentiation. The aim of this study
Glenda J Dickson et al.
Experimental hematology, 37(3), 322-333 (2009-01-23)
Hemopoietic progenitor cells express clustered homeobox (Hox) genes in a pattern characteristic of their lineage and stage of differentiation. In general, HOX expression tends to be higher in more primitive and lower in lineage-committed cells. These trends have led to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service