Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501342

Sigma-Aldrich

Anti-CARD10 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CARD-containing MAGUK protein 3, CARD10, CARMA3, Carma 3, Caspase recruitment domain-containing protein 10

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 115 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CARD10(29775)

General description

Anti-CARD10 antibody detects endogenous levels of total CARD10 protein.
CARD10 (caspase recruitment domain-containing protein 10) is an important BCL10 (B-cell CLL/lymphoma 10) interactor. It is a member of membrane-associated guanylate kinase family. It is mapped to human chromosome 22q13.1. The CARD10 protein has 1032 amino acids with an estimated molecular weight of 116 kDa. It consists of five putative functional domains, an N-terminal CARD domain, a central coiled-coil domain, and a C-terminal tripartite structure consisting of a PDZ (post synaptic density protein (PSD95), drosophila disc large tumor suppressor (Dlg1) and zonula occludens-1 protein) domain, an SH3 (SRC homology 3 domain) domain, and a GUK (guanylate kinases) domain with homology to guanylate kinase. In several adult tissues, including heart, kidney, and liver, it is expressed as a 4.4-kilobase transcript.

Immunogen

The antiserum was produced against synthesized peptide derived from human CARD10.

Immunogen Range: 481-530

Biochem/physiol Actions

CARD10 (Caspase recruitment domain-containing protein 10) helps to arrange signaling complexes at plasma membrane. It also serves as a molecular scaffold for the assembly of a B-cell CLL/lymphoma 10 (BCL10) signaling complex that activates NF-κB (nuclear factor-κB).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genome-wide association studies in Asians confirm the involvement of ATOH7 and TGFBR3, and further identify CARD10 as a novel locus influencing optic disc area.
Khor C C, et al.
Human Molecular Genetics, 20(9), 1864-1872 (2011)
Card10 is a novel caspase recruitment domain/membrane-associated guanylate kinase family member that interacts with BCL10 and activates NF-kappa B.
Wang L, et al.
The Journal of Biological Chemistry, 276(24), 21405-21409 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service