Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501317

Sigma-Aldrich

Anti-MYLIP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Inducible degrader of the low-density lipoprotein receptor, E3 ubiquitin-protein ligase MYLIP, MIR, Myosin regulatory light chain-interacting protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYLIP(29116)

General description

The MYLIP (myosin regulatory light chain interacting protein) gene is mapped to human chromosome 6p22.3. The encoded protein is an E3 ubiquitin ligase, expressed in all tissues including hippocampus and cerebellum. Anti-MYLIP antibody detects endogenous levels of total MYLIP protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MYLIP.

Immunogen Range: 161-210

Application

Anti-MYLIP antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

MYLIP (myosin regulatory light chain interacting protein) controls the LDLR (low density lipoprotein receptor) activity and stability, which might change due to fluctuation in the intracellular cholesterol levels. Mylip is associated with brain development. It induces platelet-derived growth factor (PDGF) signaling in fibroblasts. Mylip is also called as inducible degrader of the LDL-R (Idol), as it leads to receptor degradation, especially in macrophages and liver. It might serve as a target for drug development in disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dan-Qian Chen et al.
Nephrology, dialysis, transplantation : official publication of the European Dialysis and Transplant Association - European Renal Association, 32(7), 1154-1166 (2017-03-25)
The kidney plays a central role in elimination of metabolic waste products and regulation of low-molecular weight metabolites via glomerular filtration, tubular secretion and reabsorption. Disruption of these processes results in profound changes in the biochemical milieu of the body
Mylip makes an Idol turn into regulation of LDL receptor
Lindholm D, et al.
Cellular and Molecular Life Sciences, 66(21), 3399-3402 (2009)
Role of PCSK9 and IDOL in the pathogenesis of acquired LDL receptor deficiency and hypercholesterolemia in nephrotic syndrome
Liu S and Vaziri ND
Nephrology, Dialysis, and Transplantation, 29(3), 538-543 (2013)
RP1-13d10. 2 is a novel modulator of statin-induced changes in cholesterol
Mitchel K, et al.
Circulation: Genomic and Precision Medicine, 9(3), 223-230 (2016)
Qi Yu et al.
Molecular medicine reports, 18(3), 2780-2788 (2018-07-18)
Low‑density lipoprotein receptors (LDLRs) may serve a role in the diabetogenic effect of statins; however, the effects of statins on LDLR expression and its regulation in the pancreas and islets have yet to be determined. To exclude the long‑term effects

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service