Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501310

Sigma-Aldrich

Anti-GR antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GCR, GRL, GRL1, NR3C1, glucocorticoid receptor

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 85 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCR(2908)

General description

Anti-GR antibody detects endogenous levels of total GR protein.
Glucocorticoid receptor (GR) gene/NR3C1 (nuclear receptor subfamily 3 group C member 1) is located on human chromosome 5q31.

Immunogen

The antiserum was produced against synthesized peptide derived from human GR.

Immunogen Range: 201-250

Application

Anti-GR antibody has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

The glucocorticoid receptor (GR) gene/(NR3C1) is associated with asthma. It is required for glucose homeostasis, hyperinsulinemia and increased fat deposition. It also regulates cell differentiation, nervous and immune system functions.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of Hippocampal NMDA Receptors in Long-Term Emotional Responses following Muscarinic Receptor Activation
Hoeller AA, et al.
PLoS ONE, 11(1), e0147293-e0147293 (2016)
Alexandre A Hoeller et al.
PloS one, 11(1), e0147293-e0147293 (2016-01-23)
Extensive evidence indicates the influence of the cholinergic system on emotional processing. Previous findings provided new insights into the underlying mechanisms of long-term anxiety, showing that rats injected with a single systemic dose of pilocarpine--a muscarinic receptor (mAChR) agonist--displayed persistent
Glucocorticoid receptor polymorphism in obesity and glucose homeostasis
Majer-Lobodzinska A and Adamiec-Mroczek J
Advances in Clinical and Experimental Medicine : Official Organ Wroclaw Medical University, 26(1), 143-148 (2017)
Identification of polymorphisms in the human glucocorticoid receptor gene (NR3C1) in a multi-racial asthma case and control screening panel
Hawkins GA, et al.
Dna Sequence, 15(3), 167-173 (2004)
Structure of the glucocorticoid receptor (NR3C1) gene 5' untranslated region: identification, and tissue distribution of multiple new human exon 1
Turner JD and Muller CP
Journal of Molecular Endocrinology, 35(2), 283-292 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service