Skip to Content
Merck
All Photos(2)

Key Documents

SAB4501126

Sigma-Aldrich

Anti-GATA3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GATA-binding factor 3, Trans-acting T-cell specific transcription factor GATA-3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 47 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GATA3(2625)

General description

Anti-GATA3 antibody detects endogenous levels of total GATA3 protein.
GATA-3 (GATA binding protein 3) is a T-cell transcription factor. It is highly expressed in fetal and adult thymus. It is located on human chromosome 10p14.

Immunogen

The antiserum was produced against synthesized peptide derived from human GATA3.

Immunogen Range: 274-323

Biochem/physiol Actions

GATA-3 (GATA binding protein 3) plays an important role in the growth of T-cell neoplasms and early growth of T-cells. It acts as a specific marker in breast and urothelial carcinomas. GATA3 also acts as an immunophenotypic marker of acute leukemias.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T-cell transcription factor GATA-3 is an immunophenotypic marker of acute leukemias with T-cell differentiation
Dorfman DM, et al.
Human Pathology, 65, 166-174 (2017)
A genome-wide association study of Hodgkin's lymphoma identifies new susceptibility loci at 2p16.1 (REL), 8q24.21 and 10p14 (GATA3)
Enciso-Mora V, et al.
Nature Genetics (2010)
Characterisation of GATA3 expression in invasive breast cancer: differences in histological subtypes and immunohistochemically defined molecular subtypes
Shaoxian T, et al.
Journal of Clinical Pathology (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service