Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500771

Sigma-Aldrich

Anti-P300 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

E1A-associated protein p300, EP300, P300 transcription coactivator homolog

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1204


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1204

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 264 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EP300(2033)

General description

E1A binding protein p300 (p300) has a central bromodomain and three cysteine- and histidine-rich regions. The carboxy-terminal region interacts with the adenovirus oncoprotein E1A. The gene encoding this protein is localized on human chromosome 22q13. Anti-P300 Antibody detects endogenous levels of total P300 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human p300.

Immunogen Range: 55-104

Biochem/physiol Actions

E1A binding protein p300 (p300) and CBP (CREB binding protein) are highly related transcriptional coactivators. Both proteins have been identified through protein interaction assays. In addition to interacting with a variety of cellular factors and oncoproteins, loss of the wild type CBP alleles in isolated tumors suggests that CBP/p300 might serve as tumor suppressors. The ability of p300 to acetylate many transcription factors, including tumor suppressor p53, E2 factor (E2F), transcription factors II E and -F (TFIIE and -F) etc. demonstrates a novel mechanism of targeted p300 regulation of gene expression. Mutations in the gene encoding the protein have been linked to Rubinstein-Taybi syndrome (RSTS).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expanding the phenotypic spectrum in EP300-related Rubinstein-Taybi syndrome.
Solomon BD
American Journal of Medical Genetics, 167A(5), 1111-1116 (2015)
Molecular cloning and functional analysis of the adenovirus E1A-associated 300-kD protein (p300) reveals a protein with properties of a transcriptional adaptor.
Eckner R
Genes & Development, 8(8), 869-884 (1994)
Zhi Chen et al.
Molecular therapy. Nucleic acids, 21, 1100-1110 (2020-09-01)
Monocytes and macrophages are the two major cell types involved in innate immunity. Exosomes act as signaling molecules to regulate cell-to-cell communication by releasing proteins, mRNAs, microRNAs (miRNAs), and long noncoding RNAs (lncRNAs). However, it is still unclear whether monocyte-derived
Jihong Chen et al.
Scientific reports, 5, 13727-13727 (2015-09-12)
Skeletal myogenesis is a highly ordered process which specifically depends on the function of transcriptional coactivator p300. Previous studies have established that Akt/protein kinase B (PKB), a positive regulator of p300 in proliferating cells, is also important for proper skeletal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service