SAB4500690
Anti-EDG3 antibody produced in rabbit
affinity isolated antibody
Synonym(s):
Endothelial differentiation G-protein coupled receptor 3, S1P receptor Edg-3, S1P3, Sphingosine 1-phosphate receptor 3, Sphingosine 1-phosphate receptor Edg-3
Select a Size
€394.00
Select a Size
About This Item
€394.00
Recommended Products
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen 42 kDa
species reactivity
human
concentration
~1 mg/mL
technique(s)
ELISA: 1:5000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
SMILES string
O1C(C1C\C=C\CCCCCCCC(=O)OC)CCCCC
InChI
1S/C19H34O3/c1-3-4-11-14-17-18(22-17)15-12-9-7-5-6-8-10-13-16-19(20)21-2/h9,12,17-18H,3-8,10-11,13-16H2,1-2H3/b12-9+
InChI key
JTSVQVYMBXVLFI-FMIVXFBMSA-N
Gene Information
human ... S1PR3(1903)
General description
Immunogen
Immunogen Range: 1-50
Features and Benefits
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service