Skip to Content
Merck
All Photos(3)

Key Documents

SAB4500455

Sigma-Aldrich

Anti-CBP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CREB-binding protein, CREBBP

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 265 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CREBBP(1387)

General description

Anti-CBP Antibody detects endogenous levels of total CBP protein. CREB binding protein (CBP) is a histone acetyltransferase, which is part of the KAT3 family of histone acetyltransferases. The gene encoding it is localized on human chromosome 16p13.3.

Immunogen

The antiserum was produced against synthesized peptide derived from human CBP.

Immunogen Range: 1501-1550

Biochem/physiol Actions

CREB binding protein (CBP) has a role in the activation of the innate immune response, activation of histones and non-histones through acetylation. It also plays an important role in cAMP-dependent and Wnt signaling. The protein is also involved in modulating the cell cycle, DNA damage repair, ubiquitination and antigen presentation. Mutations in the CBP gene have been associated with Rubinstein-Taybi syndrome. It has also been linked with acute lymphoblastic leukemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CREBBP knockdown enhances RAS/RAF/MEK/ERK signaling in Ras pathway mutated acute lymphoblastic leukemia but does not modulate chemotherapeutic response.
Dixon ZA
Haematologica (2017)
Submicroscopic deletions at 16p13.3 in
Rubinstein-Taybi syndrome: frequency and
clinical manifestations in a North American
population
Robert Wallerstein
Journal of medical Genetics (1997)
Jin-Ran Chen et al.
The Journal of endocrinology, 239(1), 33–47-33–47 (2018-10-12)
Intrauterine or early postnatal high-fat diet (HFD) has substantial influences on adult offspring health; however, studies of HFD-induced maternal obesity on regulation of adult offspring bone formation are sparse. Here, we investigated the effects of HFD-induced maternal obesity on both

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service