Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500418

Sigma-Aldrich

Anti-GPR151 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G-protein coupled receptor PGR7, GPCR-2037, GPR151, PGR7, Probable G-protein coupled receptor 151

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 46 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR151(134391)

General description

G protein-coupled receptor 151 (GPR151) is encoded by the gene mapped to human chromosome 5. It belongs to the A type of G protein-coupled receptor (GPCR) family and it shows homology with galanin receptor 2 and 3. In mice, GPR151 transcript is highly and specifically expressed in habenular neurons.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR151.

Immunogen Range: 370-419

Application

Anti-GPR151, C-Terminal antibody produced in rabbit has been used for Western blotting.

Biochem/physiol Actions

G protein-coupled receptor 151 (GPR151) is a highly conserved protein, and acts as a potential target for psychiatric drug development, due to its expression in habenular neurons which may be critically involved in the pathophysiology of psychiatric disorders such as addiction and depression. Experimental studies reveal that GPR151 might play a neuroprotective and/or pro-regenerative role in the dorsal root ganglion (DRG) after nerve injury.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conserved expression of the GPR151 receptor in habenular axonal projections of vertebrates.
Broms J, et al.
The Journal of Comparative Neurology, 523(3), 359-380 (2015)
Nisa Roy et al.
Scientific reports, 11(1), 5549-5549 (2021-03-12)
G-protein coupled receptor 139 (GPR139) is an evolutionarily conserved orphan receptor, predominantly expressing in the habenula of vertebrate species. The habenula has recently been implicated in aversive response and its associated learning. Here, we tested the hypothesis that GPR139 signalling
Yuxiao Yao et al.
Molecular neurobiology (2023-07-21)
Histamine receptors mediate important physiological processes and take part in the pathophysiology of different brain disorders. Histamine receptor 1 (HRH1) is involved in the development of neurotransmitter systems, and its role in neurogenesis has been proposed. Altered HRH1 binding and
Targeted disruption of the orphan receptor Gpr151 does not alter pain-related behaviour despite a strong induction in dorsal root ganglion expression in a model of neuropathic pain.
Holmes FE, et al.
Molecular and Cellular Neurosciences, 78, 35-40 (2017)
Jonas Broms et al.
The Journal of comparative neurology, 525(15), 3227-3250 (2017-06-29)
GPR151 is a G-protein coupled receptor for which the endogenous ligand remains unknown. In the nervous system of vertebrates, its expression is enriched in specific diencephalic structures, where the highest levels are observed in the habenular area. The habenula has

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service