Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500306

Sigma-Aldrich

Anti-Histone H4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

H4F2, HIST, HIST1H4A, Histone H4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€347.00

€347.00


Estimated to ship on02 May 2025



Select a Size

Change View
100 μG
€347.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€347.00


Estimated to ship on02 May 2025


biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 11 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... H4-16(121504)

Related Categories

General description

Anti-Histone H4 Antibody detects endogenous levels of total Histone H4 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human Histone H4.

Immunogen Range: 13-62

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingfang Zeng et al.
The Journal of biological chemistry, 288(44), 31853-31866 (2013-09-21)
Histone deacetylase 3 (HDAC3) plays a critical role in the maintenance of endothelial integrity and other physiological processes. In this study, we demonstrated that HDAC3 undergoes unconventional splicing during stem cell differentiation. Four different splicing variants have been identified, designated
Papita Mandal et al.
The FEBS journal, 281(23), 5292-5308 (2014-09-30)
Glutamate dehydrogenase has been recently identified as a tissue-specific histone H3-specific clipping enzyme. We have previously shown that it cleaves free as well as chromatin-bound histone H3. However, the physiological significance of this enzyme is still not clear. The present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service