Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500090

Sigma-Aldrich

Anti-ADARB1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

RNA-editing deaminase 1, RNA-editing enzyme 1, dsRNA adenosine deaminase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADARB1(104)

Related Categories

General description

Adenosine deaminase, RNA specific B1 (ADARB1) is encoded by the gene mapped to human chromosome 21q22.3, a region associated with familial bipolar disorder. ADARB1 is characterized with two double-stranded RNA-binding domains and a deaminase domain involved in editing action.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADARB1.

Immunogen Range: 481-530

Biochem/physiol Actions

Adenosine deaminase, RNA specific B1 (ADARB1) enzyme plays a vital role in editing pre-mRNA of glutamate receptor B subunit. Combined effect of ADARB1 and HTR2C (5-Hydroxytryptamine Receptor 2C) variants contribute to the suicide attempt (SA) vulnerability in psychiatric patients. ADARB1 is a candidate gene for neurological diseases, such as bipolar affective disorder and epilepsy. Decrease in the activity of ADAR2 enzyme results in α-amino-3-hydroxy-5-methyl-4-isoxazolepropionic acid (AMPA) receptor-mediated death of motor neurons in mice. ADARB1 enzyme is expressed consistently in carcinomas and sarcomas, and it is upregulated in lymphomas and seminomas when compared to normal tissues.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genomics screen in transformed stem cells reveals RNASEH2A, PPAP2C, and ADARB1 as putative anticancer drug targets.
Flanagan JM, et al.
Molecular Cancer Therapeutics, 8(1), 249-260 (2009)
Sequence analysis of ADARB1 gene in patients with familial bipolar disorder.
Amore M, et al.
Journal of Affective Disorders, 81(1), 79-85 (2004)
Induced loss of ADAR2 engenders slow death of motor neurons from Q/R site-unedited GluR2.
Hideyama T, et al.
The Journal of Neuroscience, 30(36), 11917-11925 (2010)
Joint effect of ADARB1 gene, HTR2C gene and stressful life events on suicide attempt risk in patients with major psychiatric disorders.
Karanovic J, et al.
The World Journal of Biological Psychiatry : The Official Journal of the World Federation of Societies of Biological Psychiatry, 16(4), 261-271 (2015)
L Mittaz et al.
Genomics, 41(2), 210-217 (1997-04-15)
RED1 is a double-stranded RNA-specific editase characterized in the rat and is implicated in the editing of glutamate receptor subunit pre-mRNAs, particularly in the brain. Starting from human ESTs homologous to the rat RED1 sequence, we have characterized two forms

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service