Skip to Content
Merck
All Photos(3)

Documents

SAB4500087

Sigma-Aldrich

Anti-Tubulin α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

α-tubulin 1, TBA1, TBA1A, TBA4A, Testis-specific α- tubulin

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 50 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TUBA1A(7846)

General description

Testis-specific α-tubulin is an α tubulin isotype expressed in testis wherein is works with β tubulin to regulate the structural and functional development of axonemes.

Specificity

Rabbit polyclonal anti-Tubulin α, C-Terminal antibody reacts with rat, mouse, human testis-specific α-tubulins.

Immunogen

The antiserum was produced against synthesized peptide derived from human Tubulin alpha.

Immunogen Range: 402-451

Application

Rabbit polyclonal anti-Tubulin α, C-Terminal antibody is used to tag testis-specific α-tubulin (TBA1A) for detection and quantitation by immunocytochemical and immunohistochemical (IHC) techniques such as immunoblotting, immunoprecipitation, and immunofluorescence. It is used as a probe to determine the presence and roles of testis-specific α-tubulin (TBA1A) in sperm axoneme formation and function.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shunqin Zhu et al.
Experimental and therapeutic medicine, 17(5), 3883-3890 (2019-04-23)
The neural crest is a transient embryonic tissue that initially generates neural crest stem cells, which then migrate throughout the body to give rise to a variety of mature tissues. It was proposed that the fate of neural crest cells
Yi Yu et al.
Oxidative medicine and cellular longevity, 2023, 4654083-4654083 (2023-01-20)
Endothelial cell (EC) senescence characterized by an irreversible growth arrest leading to endothelial dysfunction has been implicated in vascular aging and aging-associated cardiovascular diseases. Autophagy plays a crucial role in the modulation of cellular senescence. Our previous showed that myosin
Mohamed Jemaà et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 54(2), 303-320 (2020-04-08)
Chromosomal instability is a well-known factor in the progression of different types of cancer, including colorectal cancer. Chromosomal instability results in severely rearranged karyotypes and aneuploidy. Tetraploidy constitutes an intermediate phase during the polyploidy/aneuploidy cascade in oncogenesis, and tetraploid cells
Elham Taha et al.
Current biology : CB, 30(18), 3507-3521 (2020-07-25)
Levels of adult neurogenesis in the dentate gyrus (DG) of the hippocampus are correlated with unique cognitive functions. However, the molecular pathways controlling it are poorly understood. Here, we found that the known physiological ways to enhance neurogenesis converged on
Hye-Yeon Lee et al.
Aging and disease, 14(4), 1407-1424 (2023-05-10)
The term probiotic refers to bacteria that provide a beneficial effect to the host. Limosilactobacillus reuteri (Lactobacillus reuteri) is a probiotic isolated from human breast milk. Although L. reuteri has antimicrobial and anti-inflammatory activities occasionally linked to anti-aging effects, there

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service