Skip to Content
Merck
All Photos(2)

Key Documents

SAB4500068

Sigma-Aldrich

Anti-p27 KIP1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CDKN1B, CDN1B, Cyclin-dependent kinase inhibitor 1B, Cyclin-dependent kinase inhibitor p27, KIP1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€463.00

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€463.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€463.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 22 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDKN1B(1027)

General description

Anti-p27 KIP1 Antibody detects endogenous levels of total p27 KIP1 protein.
Cyclin-dependent kinase inhibitor 1B (CDKN1B or p27 KIP1) is a member of CDK inhibitor (CKI). The CDKN1B gene is mapped to human chromosome 12p13.1. The prime domains in CDKN1B include the nuclear export signal (NES) and the nuclear localization signal (NLS) and the CDK binding domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human p27 Kip1.

Immunogen Range: 1-50

Application

Anti-p27 KIP1 antibody produced in rabbit has been used in western blotting[1] (1:500).[2]

Biochem/physiol Actions

Cyclin-dependent kinase inhibitor 1B (CDKN1B) negatively regulates cell cycle events by binding to cyclin-dependent kinase. It is a tumor suppressor gene correlated with the pathophysiology of tumors. Germline mutations in CDKN1B are implicated in multiple endocrine neoplasia (MEN) syndrome and MEN-like syndrome (MENX). It is a potent biomarker in acute myeloid leukemia.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martina Cusan et al.
Frontiers in endocrinology, 9, 393-393 (2018-08-02)
The CDKN1B gene encodes for the p27Kip1 protein, firstly characterized as a cyclin dependent kinase (CDK)-inhibitor. Germline CDKN1B pathogenic variants have been described in hereditary tumors, such as multiple endocrine neoplasia (MEN)-like syndromes and familial prostate cancer. Despite its central
Martina Kluth et al.
Oncotarget, 6(29), 27966-27979 (2015-08-22)
Deletion of 12p is a recurrent alteration in prostate cancer, but the prevalence and clinical consequences of this alteration have not been studied in detail. Dual labeling fluorescence in situ hybridization using probes for 12p13 (CDKN1B; p27) and centromere 12
R McKay
Current opinion in neurobiology, 2(5), 582-585 (1992-10-01)
Several recent experiments have shown that immortal neural and muscle cells retain the specific temporal and spatial properties of their precursor cells in the animal. In addition, the immortal cells retain the ability to differentiate in vitro and, after transplantation
Natalia S Pellegata et al.
Proceedings of the National Academy of Sciences of the United States of America, 103(42), 15558-15563 (2006-10-13)
MENX is a recessive multiple endocrine neoplasia-like syndrome in the rat. The tumor spectrum in MENX overlaps those of human multiple endocrine neoplasia (MEN) types 1 and 2. We mapped the MenX locus to the distal part of rat chromosome
Gian Luca Rampioni Vinciguerra et al.
Cell division, 14, 2-2 (2019-04-13)
The p27kip1 protein, mainly known as a negative regulator of cell proliferation, has also been involved in the control of other cellular processes, including the regulation of cytoskeleton dynamics. Notably, these two functions involve distinct protein domains, residing in the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service