Skip to Content
Merck
All Photos(1)

Key Documents

SAB4500050

Sigma-Aldrich

Anti-CDK1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cell division protein kinase 2, kinase Cdk2, p33 protein kinase

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€394.00

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€394.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€394.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 33 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDK2(1017)

General description

Anti-CDK1 Antibody detects endogenous levels of total CDK1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CDK1/CDC2.

Immunogen Range: 1-50

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Praveen Kumar Reddy Nagireddy et al.
ACS omega, 4(21), 19382-19398 (2019-11-26)
Noscapine, a phthalide isoquinoline alkaloid isolated from the opium poppy Papaver somniferum, is traditionally being used as an anticough drug. With a safe in vitro toxicological profile, noscapine and its analogues have been explored to show microtubule-regulating properties and anticancer
Xiaohan Qu et al.
Journal of translational medicine, 13, 301-301 (2015-09-17)
PIWI proteins have important roles in tumorigenesis due to their interaction with piRNAs. Recent studies suggest that PIWI proteins affect prognosis of various cancers. In the present study, PIWI genes expression was assayed in non-small cell lung cancer (NSCLC). To

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service