Skip to Content
Merck
All Photos(1)

Key Documents

SAB4301142

Sigma-Aldrich

Anti-BSA antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ALB, ALBU, Anti-Bsa Antibody, Anti-Bsa Antibody - Anti-BSA antibody produced in rabbit, albumin, cell growth inhibiting protein 42

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€329.00

€329.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μL
€329.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€329.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

69 kDa

species reactivity

bovine

concentration

0.7 mg/mL

technique(s)

western blot: 1:1000-1:5000 (Cell Lysate)

isotype

IgG

accession no.

NP_851335.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

bovine ... ALB(280717)

General description

Bovine serum albumin (BSA) is a single polypeptide chain consisting of about 583 amino acid residues and no carbohydrates. It is a 66.4 kDa, water-soluble protein. BSA is characterized with three homologous domains formed by six α-helices. Depending on pH, it undergoes reversible conformational isomerization. The native structure of the protein becomes reactive and flexible on heating.

Specificity

The antibody detects endogenous levels of total BSA protein.

Immunogen

Fusion protein corresponding to a region derived from internal residues of human Bovine serum albumin

Biochem/physiol Actions

Bovine serum albumin (BSA) has been used as a model protein in various studies, such as immunodiagnostic procedures, cell culture media and clinical chemistry and relevant in foods containing bovine milk or meat. Serum albumins play a dominant role in drug disposition and efficacy by maintaining osmotic blood pressure.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Emily A Willmann et al.
BMC immunology, 21(1), 61-61 (2020-12-16)
C1q is a soluble pattern recognition protein that regulates multiple leukocyte functions, and deficiency in C1q results in autoimmunity. C1q stimulates enhanced phagocytic function through multiple mechanisms including the rapid enhancement of Fcγ receptor (FcγR) -mediated phagocytosis. The molecular mechanism
Study of the interaction between monoammonium glycyrrhizinate and bovine serum albumin.
Hu Y J, et al.
Journal of Pharmaceutical and Biomedical Analysis, 36(4), 915-919 (2004)
Jonas B Lang et al.
Frontiers in oncology, 12, 949261-949261 (2022-10-21)
Cellular crosstalk between hematopoietic stem/progenitor cells and the bone marrow (BM) niche is vital for the development and maintenance of myeloid malignancies. These compartments can communicate via bidirectional transfer of extracellular vesicles (EVs). EV trafficking in acute myeloid leukemia (AML)
Kinetics of Thermal Denaturation and Aggregation of Bovine Serum Albumin.
Borzova VA
PLoS ONE, 11(4), e0153495-e0153495 (2016)
Ine Rombouts et al.
Scientific reports, 5, 12210-12210 (2015-07-21)
Thermolysin hydrolyzates of freshly isolated, extensively stored (6 years, 6 °C, dry) and heated (60 min, 90 °C, in excess water) bovine serum albumin (BSA) samples were analyzed with liquid chromatography (LC) electrospray ionization (ESI) tandem mass spectrometry (MS/MS) using alternating electron-transfer

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service