Skip to Content
Merck
All Photos(2)

Documents

SAB4300568

Sigma-Aldrich

Anti-FAM129B (Ab-712) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C9orf88 antibody produced in rabbit, Anti-DKFZP434H0820 antibody produced in rabbit, Anti-FLJ13518 antibody produced in rabbit, Anti-FLJ22151 antibody produced in rabbit, Anti-family with sequence similarity 129, member B antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~82 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(Q-V-S-S-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FAM129B(64855)

Immunogen

Peptide sequence around aa. 710-714 (Q-V-S-S-P), according to the protein FAM129B.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

a maternal imprinting gene. May possess growth suppressor activity. Two major alternatively spliced isoforms have been described. forms. One form, Meg3-proximal ( Meg3p), contains exons 1-3. The second form, Meg3-distal ( Meg3d) did not contain exons 1-3 and was present in oocytes and in 1- and 2-cell embryos.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service