Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300512

Sigma-Aldrich

Anti-NF2 (Ab-518) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-ACN antibody produced in rabbit, Anti-BANF antibody produced in rabbit, Anti-SCH antibody produced in rabbit, Anti-neurofibromin 2 (merlin) antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~69 kDa

species reactivity

mouse, human, rat

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(R-L-S-M-E)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NF2(4771)

General description

Neurofibromin 2 (NF2) encodes the protein Merlin, which belongs to Ezrin/Radixin/Moesin (ERM) family of proteins. NF2 is a cytoskeletal protein involved in stabilization of microtubules. NF2 has four-point-one, ezrin, radixin, moesin (FERM) domain in the N-terminal, a coiled-coil domain, and does not contain C-terminal actin-binding motif like other ERM proteins. In human chromosome, the gene NF2 is localized on 22q12.2

Immunogen

Peptide sequence around aa. 516-520 (R-L-S-M-E), according to the protein NF2.

Biochem/physiol Actions

Neurofibromin 2 (NF2) is a tumour suppressor. NF2 translocates to nucleus and binds to CRL4DCAF1 (Cullin Ring E3 ubiquitin ligase cullin 4 (CRL4) DDB1 (DNA damage binding protein 1) and CUL4 (cullin 4) associated factor 1 (DCAF1)) and suppresses tumour growth. The gene NF2 is inactivated in the familial neurofibromatosis type 2 cancer syndrome. Mutations of NF2 is associated with multiple tumours like Schwannomas, meningiomas, and malignant mesotheliomas. NF2 regulates Hippo signalling pathway and further activates yes associated protein 1 (YAP-1) and modulates cell growth in neurofibromatosis 2. Molecular modifications in NF2 gene leads to liver cancer through the involvement of Hippo pathway of pathogenesis. Polymorphism in NF2 is related to medulloblastoma.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Probable regulator of the Hippo/SWH (Sav/Wts/Hpo) signaling pathway, a signaling pathway that plays a pivotal role in tumor suppression by restricting proliferation and promoting apoptosis. Along with WWC1 can synergistically induce the phosphorylation of LATS1 and LATS2 and can probably function in the regulation of the Hippo/SWH (Sav/Wts/Hpo) signaling pathway. May act as a membrane stabilizing protein. May inhibit PI3 kinase by binding to AGAP2 and impairing its stimulating activity.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NF2/merlin inactivation and potential therapeutic targets in mesothelioma
Sato T and Yoshitaka S
International Journal of Molecular Sciences, 19(4), 988-988 (2018)
Microdeletion del (22)(q12. 2) encompassing the facial development-associated gene, MN1 (meningioma 1) in a child with Pierre-Robin sequence (including cleft palate) and neurofibromatosis 2 (NF2): a case report and review of the literature
Davidson TB, et al.
BMC Medical Genetics, 13, 19-19 (2012)
The association of NF2 (neurofibromin 2) gene polymorphism and the risk of medulloblastomas
Zhao C, et al.
Neurological Sciences, 39, 1175-1183 (2018)
Merlin/NF2 suppresses tumorigenesis by inhibiting the E3 ubiquitin ligase CRL4 DCAF1 in the nucleus
Li W, et al.
Cell, 140(4), 477-490 (2010)
Molecular alterations of the NF2 gene in hepatocellular carcinoma and intrahepatic cholangiocarcinoma
Zhang N, et al.
Oncology Reports, 38(6), 3650-3658 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service