Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300428

Sigma-Aldrich

Anti-FOXO1 (Ab-319) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-FKH1 antibody produced in rabbit, Anti-FKHR antibody produced in rabbit, Anti-FOXO1A antibody produced in rabbit, Anti-forkhead box O1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

78-82 kDa

species reactivity

mouse, human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(T-S-S-N-A)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FOXO1(2308)

General description

FOXO1 (forkhead box O1) is a transcription factor , that belongs to the Foxo subfamily. It is located on human chromosome 13q14.

Immunogen

Peptide sequence around aa. 317-321 (T-S-S-N-A), according to the protein FOXO1.

Biochem/physiol Actions

FOXO1 (forkhead box O1) blocks the differentiation of cells in fetal pancreatic progenitor cells. It controls the differentiation of adipocytes.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Transcription factor.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Forkhead Protein FoxO1 Acts as a Repressor to Inhibit Cell Differentiation in Human Fetal Pancreatic Progenitor Cells
Jiang Z, et al.
Journal of Diabetes Research (2017)
The forkhead transcription factor Foxo1 regulates adipocyte differentiation
Nakae J, et al.
Developmental Cell, 4(1), 119-129 (2003)
Dense pattern of embryonal rhabdomyosarcoma, a lesion easily confused with alveolar rhabdomyosarcoma: a report from the Soft Tissue Sarcoma Committee of the Children's Oncology Group
Rudzinski ER, et al.
American Journal of Clinical Pathology, 140(1), 82-90 (2013)
The forkhead transcription factor Foxo1 links insulin signaling to Pdx1 regulation of pancreatic beta cell growth
Kitamura T, et al.
The Journal of Clinical Investigation, 110(12), 1839-1847 (2002)
Zhixia Fan et al.
International journal of clinical and experimental pathology, 13(4), 799-809 (2020-05-02)
Psoriasis is an immune-mediated skin disease with abnormal T cells. Regulatory T cells (Treg) are a kind of cell group with immunosuppressive effects. This study aimed to explore the role of Treg cells in the pathogenesis of psoriasis and its

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service