Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300389

Sigma-Aldrich

Anti-CAV1 (Ab-14) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-BSCL3 antibody produced in rabbit, Anti-CAV antibody produced in rabbit, Anti-CGL3 antibody produced in rabbit, Anti-MSTP085 antibody produced in rabbit, Anti-caveolin 1, caveolae protein, 22kDa antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€342.00

€342.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1602


Select a Size

Change View
100 μG
€342.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€342.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)
A recombinant, preservative-free antibody is available for your target. Try ZRB1602

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~24 kDa

species reactivity

mouse, rat, human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(H-L-Y-T-V)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CAV1(857)

General description

Caveolin-1 (CAV1) is a multifunctional scaffolding protein , that has three exons. It is located on human chromosome 7q31.

Immunogen

Peptide sequence around aa. 12-16 (H-L-Y-T-V), according to the protein CAV1.

Biochem/physiol Actions

Caveolin-1 is expected to play a major role in the growth and evolution of cancer. It participates in glucose metabolism. It may also participate in angiogenesis. Cav-1 helps to control the expression of glucose transporters.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

The scaffolding protein encoded by Caveolin-1 is the main component of the caveolae plasma membranes found in most cell types. The protein links integrin subunits to the tyrosine kinase FYN, an initiating step in coupling integrins to the Ras-ERK pathway and promoting cell cycle progression. The gene is a tumor suppressor gene candidate and a negative regulator of the Ras-p42/44 MAP kinase cascade. CAV1 and CAV2 are located next to each other on chromosome 7 and express colocalizing proteins that form a stable hetero-oligomeric complex. By using alternative initiation codons in the same reading frame, two isoforms (alpha and beta) are encoded by a single transcript from this gene.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genes encoding human caveolin-1 and -2 are co-localized to the D7S522 locus (7q31.1), a known fragile site (FRA7G) that is frequently deleted in human cancers
Engelman JA, et al.
Febs Letters, 436(3), 403-410 (1988)
GDM-Induced Macrosomia Is Reversed by Cav-1 via AMPK-Mediated Fatty Acid Transport and GLUT1-Mediated Glucose Transport in Placenta
Yao G, et al.
PLoS ONE, 12(1) (2017)
Aberrant CpG Island Shore Region Methylation of CAV1 Is Associated with Tumor Progression and Poor Prognosis in Gastric Cardia Adenocarcinoma
Guo YL, et al.
Archives of Medical Research, 47(6), 460-470 (2016)
caveolin-1 expression in prostatic hyperplasia, high grade prostatic intraepithelial hyperplasia and prostatic carcinoma and its correlation with microvessel density
Mohammed DA and Helal DS
Journal of the Egyptian National Cancer Institute, 29(1), 25-31 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service