Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300370

Sigma-Aldrich

ANTI-TAU (MAPT) Antibody

rabbit polyclonal

Synonym(s):

Anti-DDPAC antibody produced in rabbit, Anti-FLJ31424 antibody produced in rabbit, Anti-FTDP-17 antibody produced in rabbit, Anti-MAPTL antibody produced in rabbit, Anti-microtubule-associated protein tau antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€302.00

€302.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
€302.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€302.00


Ships within 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Product Name

Anti-MAPT (Ab-356) antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~48 kDa
~62 kDa
~78 kDa

species reactivity

human, mouse, rat

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200

isotype

IgG

immunogen sequence

(I-G-S-L-D)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)

Related Categories

General description

MAPT (microtubule associated protein tau) is located on human chromosome 17q21.3. It is upregulated in neurons but is most prominent in axons.

Immunogen

Peptide sequence around aa. 354-358 (I-G-S-L-D), according to the protein MAPT.

Biochem/physiol Actions

Removal of MAPT results in developmental delay and learning disability. It participates in the pathology of Alzheimer′s disease (AD). It helps in the assembly and maintenance of microtubule structure.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Promotes microtubule assembly and stability, and might be involved in the establishment and maintenance of neuronal polarity. The C-terminus binds axonal microtubules while the N-terminus binds neural plasma membrane components, suggesting that tau functions as a linker protein between both. Axonal polarity is predetermined by tau localization (in the neuronal cell) in the domain of the cell body defined by the centrosome. The short isoforms allow plasticity of the cytoskeleton whereas the longer isoforms may preferentially play a role in its stabilization.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Microdeletion encompassing MAPT at chromosome 17q21.3 is associated with developmental delay and learning disability
Shaw-Smith C
Nature Genetics, 38(9), 1032-1037 (2006)
Linkage disequilibrium and association of MAPT H1 in Parkinson disease
Skipper L, et al.
American Journal of Human Genetics, 75(4), 669-677 (2004)
The H1c haplotype at the MAPT locus is associated with Alzheimer's disease
Myers AJ, et al.
Human Molecular Genetics, 14(16), 2399-2404 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service