Skip to Content
Merck
All Photos(3)

Key Documents

SAB4300282

Sigma-Aldrich

Anti-phospho-SYK (pTyr323) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp313N1010 antibody produced in rabbit, Anti-FLJ25043 antibody produced in rabbit, Anti-FLJ37489 antibody produced in rabbit, Anti-spleen tyrosine kinase antibody produced in rabbit

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€493.00

€493.00


Check Cart for Availability


Select a Size

Change View
100 μG
€493.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€493.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~72 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(N-P-YP-E-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr323)

Gene Information

human ... SYK(6850)

General description

Spleen tyrosine kinase (SYK) is a member of the non-receptor tyrosine kinase family. It is usually expressed in epithelial breast tissues and hematopoietic cells. The gene encoding it is localized on human chromosome 9q22.

Immunogen

Peptide sequence around phosphorylation site of tyrosine 323 (N-P-Y(p)-E- P), according to the protein SYK.

Biochem/physiol Actions

Spleen tyrosine kinase (SYK) is a tumor suppressor gene that helps to prevent the development and metastasis of breast cancer cells. It participates in B-cell receptor signaling. The protein helps to phosphorylate the α-tubulin subunits of microtubules. SYK aids in the coupling of receptors for antigen-antibody complexes. It also modulates cell proliferation and differentiation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Syk Is Recruited to Stress Granules and Promotes Their Clearance through Autophagy.
Krisenko MO
The Journal of Biological Chemistry (2015)
Syk: a new player in the field of breast cancer
Zoe A
Breast Cancer Research (2000)
Inhibition of Spleen Tyrosine Kinase Potentiates Paclitaxel-Induced Cytotoxicity in Ovarian Cancer Cells by Stabilizing Microtubules.
Yu Y
Cancer Cell (2015)
Novel SYK gene variations and changes in binding sites of miRs in breast cancer patients.
Faryal R
Cancer Biomarkers : Section A of Disease Markers (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service