Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200853

Sigma-Aldrich

Anti-PBP2a of MRSA antibody, Mouse monoclonal

clone 38, purified from hybridoma cell culture

Synonym(s):

Anti-Beta-lactam-inducible penicillin-binding protein 2a

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€153.00
100 μL
€387.00

€153.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€153.00
100 μL
€387.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

€153.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

38, monoclonal

form

liquid

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.5-1 μg/mL using whole recombinant PBP2a protein

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Infections caused by methicillin-resistant Staphylococcus aureus (MRSA) are a major concern cause for clinicians, presenting high mortality and morbidity rates compared to infections caused by methicillin-sensitive Staphylococci.2

Specificity

Monoclonal Anti-PBP2a of MRSA antibody specifically recognizes PBP2a protein from Methicillin-resistant Staphylococcus aureus (MRSA). The antibody reacts with both denaturated recombinant PBP2a protein and bacteria lysates in Immunoblot.1 The antibody recognizes PBP2a in MRSA cells in both the exponential and stationary phases, however no cross reaction with methicillin-sensitive Staphylococcus aureus (PBP2a negative) and E. coli were seen.1

Application

The antibody may be used in various immunochemical techniques including Immunoblotting1 (~76 kDa), ELISA1, Surface Plasmon Resonance (SPR)1 and Flow cytometry1.

Biochem/physiol Actions

Resistance of S. aureus to methicillin and all beta-lactam antibiotics is due to the activity of PBP2a protein (i.e., penicillin binding protein2a) that is located in the cell wall of the resistant strains.3-4 PBP2a is encoded by mecA gene and belongs to a family of penicillin binding proteins (PBPs). PBPs are membrane-bound enzymes that catalyze the transpeptidation reaction necessary for cross-linkage of peptidoglycan chains.5-6 Contrary to all PBPs, PBP2a has low affinity for all ß-lactam antibiotics.7 Therefore, in the presence of ß-lactam antibiotics, while normal PBPs are blocked, the PBP2a is able to continue catalyze the transpeptidation reactions enabling cell wall synthesis.4 PBP2a protein is suggested to be a promising candidate for anti-MRSA vaccine development.8

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.Antibody Concentration: ~ 1.0 mg/mL

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

1–2 of 2 Questions  
  1. Does that work for immunofluorescence?

    1 answer
    1. This product has not been tested specifically for immunofluorescence applications. However, as per the product datasheet, the antibody SAB4200853 may be used in various immunochemical techniques including Immunoblotting, Surface Plasmon Resonance, and Flow cytometry. To obtain the best results in different techniques and preparations, it is recommended to determine optimal working concentration by titration test.

      Please see the link below to review the product datasheet:
      https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/140/775/sab4200853-dat.pdf

      Helpful?

  2. What would be a good PBP control protein of this class in methicillin-sensitive cultures?

    1 answer
    1. The following paper shows how the antibody was characterized, and may contain useful information for additional controls: https://www.liebertpub.com/doi/10.1089/mab.2015.0013?url_ver=Z39.88-2003&rfr_id=ori%3Arid%3Acrossref.org&rfr_dat=cr_pub++0pubmed.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service