Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200843

Sigma-Aldrich

Anti-BCOR antibody, Mouse monoclonal

clone BR-4, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€107.00
100 μL
€325.00

€107.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€107.00
100 μL
€325.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€107.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

clone

BR-4

form

liquid

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using human HeLa nuclear extract
immunofluorescence: 2-4 μg/mL using human HeLa cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

BCOR encodes a ubiquitously expressed protein which is involved in BCL6-mediated transcriptional repression.1 Originally it was identified as a novel BCL6 corepressor via its interaction with the site-specific DNA-binding transcription factor BCL6, whose aberrant expression drives formation of diffuse large B-cell lymphomas.

Specificity

Monoclonal Anti-BCOR antibody specifically recognizes human BCOR protein.

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (∼190 kDa) and Immunofluorescence.

Biochem/physiol Actions

NCOR and SMRT, was recently demonstrated to occupy the majority of its DNA binding sites independently from BCL6, reflecting its multiple interaction partners. BCOR also forms a Polycomb repressor complex 1 (PRC1)-like complex with PCGF1, KDM2B, RING1, SKP1, RYBP, and RNF2, and can bind to both class I and II histone deacetylases.1 BCOR is specifically and strongly expressed during embryonic development where it plays a critical role in transcriptional regulation, as BCOR mutations are associated with the X-linked inherited diseases Lenz microphthalmia and Oculofaciocardiodental syndrome (OFCD).

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. What is the immunogen sequence?

    1 answer
    1. The immunogen for the BR-4 clone is a recombinant peptide from the N-terminal region of the human BCOR protein (GeneID 54880).

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service