Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200838

Sigma-Aldrich

Anti-Rubicon antibody, Mouse monoclonal

clone RB-145, purified from hybridoma cell culture

Synonym(s):

Beclin-1 associated RUN domain containing protein, KIAA0226, RUBCN, Run domain Beclin-1-interacting and cysteine-rich domain-containing protein, SCAR15

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€149.00
100 μL
€400.00

€149.00


Estimated to ship on30 April 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€149.00
100 μL
€400.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€149.00


Estimated to ship on30 April 2025


Request a Bulk Order

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

RB-145, monoclonal

form

liquid

species reactivity

human

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using human A431 whole cell extract.
immunofluorescence: 2-4 μg/mL using human HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RUBCN(9711)

General description

Run domain Beclin-1-interacting and cysteine-rich domain-containing protein (Rubicon) also known as Beclin-1 associated RUN domain containing protein (Baron) or KIAA0226, is a negative regulator of endosomes maturation and endocytic trafficking.1-2 Rubicon knockdown studies showed enhanced autophagosome maturation and endocytosis.1 It also have a critical role in the LC3-associated phagocytosis (LAP) a noncanonical autophagy process which can be induced through the activation of extracellular receptor by different stimulators including pathogens, immune complexes and dying cells.4-5

Specificity

Monoclonal Anti-Rubicon antibody specifically recognizes Rubicon from human origin.

Immunogen

synthetic peptide corresponding to the N-terminal region of human Rubicon, conjugated to KLH

Application

The antibody may be used in various immunochemical techniques including Immunoblotting and Immunofluorescence. Detection of the Rubicon band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

The mammalian class III phosphatidylinositol 3-kinase (PtdIns3K) complex consists of three core proteins, catalytic VPS34, the adaptor VPS15 (p150) and the recruiter Beclin-1 (ATG6); these endocytosis regulators were reported to stably bind Beclin-1 ATG14, UVRAG and Rubicon.1-3 The complex Rubicon-UVRAG–Beclin-1/PtdIns3K can be found both in early and in late endosomes or lysosomes. Rubicon was described as the mediator for the recruitment of the UVRAG-Beclin-1-VPS34 complex and of ATG7 and LC3-II LAPosome.4-5 Furthermore, Rubicon has been identified as a key modulator of the inflammatory response and viral replication, during several viral infections such as hepatitis B virus (HBV). Rubicon expression is induced in negative regulation of the innate immune response resulting in enhances viral replication and possibly supporting viral immune evasion mechanism.4

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service