Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200832

Sigma-Aldrich

Anti-Porphyromonas gingivalis antibody produced in rabbit

IgG fraction of antiserum

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€152.00
100 μL
€393.00

€152.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€152.00
100 μL
€393.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€152.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

IgG fraction of antiserum

description

Research area: Microbiome

form

buffered aqueous solution

species reactivity

Porphyromonas gingivalis (Lipopolysaccharides (LPS) from whole extract)

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1:50,000-1:100,000 using dead Porphyromonas gingivalis bacteria
indirect ELISA: 1:20,000-1:40,000 using Porphyromonas gingivalis LPS for coating.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Related Categories

General description

Porphyromonas gingivalis is a nonmotile, Gram-negative, rod-shaped, anaerobic pathogenic bacterium, belongs to the Porphyromonadaceae family, phylum Bacteroidetes class.1 P. gingivalis is involved in a number of pathogenic mechanisms including tissue colonization and destruction, as well as host defense activation. This bacterium is known for the ability to colonize at the oral cavity which consider a major public health issue. It can also be found in lung, liver, or splenic abscesses and the upper gastrointestinal tract and is related to atherosclerotic plaque disease, Cardiovascular diseases, Adverse pregnancy outcomes, Rheumatoid arthritis (RA), Meningitis or brain abscesses and Alzheimer′s disease. P. gingivalis ability to aggregate in biofilms is considered a significant health risk by elevated resistance to host defense mechanisms and decreased susceptibility to conventional antimicrobials agents. P. gingivalis have several potential secreted and membranal virulence factors such as Gingipains proteolytic complex (Rgp and Kgp) which degrade various host proteins, Fimbriae (FimA and Mfa1) responsible for attachment of bacterial cells to host cell surfaces and Haemagglutinins (HagA) involved in adhesion and invasion of host cells.

Application

Anti-Porphyromonas gingivalis antibody recognizes P. gingivalis whole extract and P. gingivalis Lipopolysaccharides (LPS), the antibody has no cross reactivity with P. mirabilis or E. coli. The antibody may be used in various immunochemical techniques including Immunoblotting and ELISA. Detection of the P. gingivalis bands by Immunoblotting is specifically inhibited by the immunogen.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Disclaimer

This product is for R&D use only, not for drug, household, or other uses.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alice Harding et al.
Frontiers in aging neuroscience, 9, 398-398 (2017-12-19)
Longitudinal monitoring of patients suggests a causal link between chronic periodontitis and the development of Alzheimer's disease (AD). However, the explanation of how periodontitis can lead to dementia remains unclear. A working hypothesis links extrinsic inflammation as a secondary cause

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service