Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200830

Sigma-Aldrich

Anti-Staphylococcal Enterotoxin A-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Enterotoxin type A, SEA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

lyophilized powder

mol wt

25-30 kDa

species reactivity

Staphylococcus aureus

concentration

~4 mg/mL

technique(s)

direct ELISA: 1:2,000-1:4,000 (recommended using 1 μg/ml Staphylococcal Enterotoxin A for coating.)

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Staphylococcus aureus ... sea(22112712)

General description

Staphylococcus aureus is a Gram positive bacterium, that cause diseases in human population including food poisoning and toxic shock syndrome. S. aureus expresses several secreted virulence factors, such as various enzymes, cytotoxins, exotoxins, and exfoliative toxins. Exotoxins include more than 20 serologically classified staphylococcal enterotoxins (SEs), the best characterize are the SEs A, B through V and toxic shock syndrome toxin-1 (TSST-1), these enterotoxins are similar in activity, sequence, structure and molecular weight (~25-30 kDa).
Mainly SEA but also SEB are known as superantigens due to their ability to bind class II MHC molecules on antigen presenting cells and stimulate extensive T-cells activation followed by massive cytokine release leading to an acute toxic shock.
SE proteins have a significant resistance to heat and acid, it is also resistant to gastrointestinal proteases including pepsin, trypsin, rennin and papain. Thus, killing the S. aureus bacteria may not be sufficient to eliminate these superantigens risk from causing food poisoning.
As described in various researches, detection of SEA contamination is a major challenge in the food industry, including milk and cooked food products.
SEA high affinity to MHC class II receptors on antigen presenting cells and tissues, followed by the excessive systemic immune activation, was applied in attempts to use bispecific antibodies (BsAbs) genetically fused to SEA as a T-cell stimulator. The SEA activation resulted in a non-specific activation of MHC class II-positive tissues which causes side-effects. Mutated SEA, D227A was developed to reduce the adverse effects for directing activated T-cells toward tumor cells.

Immunogen

Purified toxin from Staphylococcus aureus, (This toxin is coded by a bacteriophage).

Application

The antibody may be used in various immunochemical techniques including ELISA.

Physical form

Supplied as a lyophilized powder.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service