Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200826

Sigma-Aldrich

Anti- Cholera Toxin-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Cholera enterotoxin subunit B (ctxB), Cholera enterotoxin, A chain (ctxA)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

peroxidase conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

lyophilized powder

mol wt

subunit mol wt ~12 kDa (non-toxic CTxB )
subunit mol wt ~27 kDa (the toxic CTxA )

species reactivity

Vibrio cholerae

concentration

~4 mg/mL

technique(s)

direct ELISA: 1:8,000-1:16,000 using 5 μg/ml Cholera Toxin for coating

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Vibrio cholerae ... ctxA(57740120) , ctxB(57740119)

General description

Cholera toxin (CTx) also known as choleragen, is an enterotoxin produced by the Gram-negative bacterium Vibrio cholerae that naturally habitat in fresh or saltwater environments. CTx is composed of two subunits, the toxic CTxA (~27 kDa) and non-toxic CTxB (~12 kDa) assembled with the stoichiometry AB5. The V. cholerae virulence factors CtxA and CtxB are located at the CTX phage genome integrated within the bacterial chromosome.

Specificity

Anti-Cholera Toxin-peroxidase antibody specifically recognizes Cholera Toxin and has no cross reactivity with Staphylococcal Enterotoxin A (SEA), Staphylococcal Enterotoxin B (SEB) or Pseudomonas Exotoxin A

Immunogen

purified toxin from Vibrio cholera

Application

Anti- Cholera Toxin-Peroxidase antibody produced in rabbit has been used in direct enzyme linked immunosorbent assay.

Biochem/physiol Actions

The A1 fragment of the cholera toxin (CTx)A subunit is translocated through the membrane of the host cell, where it catalyses the ADP-ribosylation of the α-subunit of Gs (Gsa), a regulatory component of the adenylate cyclase complex. The resulting increased level of cyclic AMP promotes a wide variety of actions, including the secretion of chloride ions in the case of intestinal epithelial cells. Most of the V. cholerae species do not cause any disease in human, but few including serotypes O1 and O139 can cause cholera pandemics. Antibodies specific for cholera toxin may be used in studies of structural and functional aspects of toxin membrane interactions and for the detection of CTxB, when used, for example, as an adjuvant when injected mucosally together with the desired antigen.

Physical form

Supplied as a lyophilized powder

Storage and Stability

Store the lyophilized product at 2–8 °C. For extended storage after reconstitution, keep at –20 °C in working aliquots. Avoid repeated freeze-thaw cycles. For continuous use after reconstitution, keep at 2–8 C for up to 1 month. Solutions at working dilution should be discarded if not used within 12 hours.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Intestinal epithelial responses to enteric pathogens: effects on the tight junction barrier, ion transport, and inflammation
Berkes J, et al.
Gut, 52, 439-439 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service