Skip to Content
Merck
All Photos(5)

Key Documents

SAB4200814

Sigma-Aldrich

Anti-TIMP2 antibody, Mouse monoclonal

enhanced validation

clone TM8, purified from hybridoma cell culture

Synonym(s):

CSC-21K, Metalloproteinase inhibitor 2, Tissue inhibitor of metalloproteinases 2

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€129.00
100 μL
€343.00

€129.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€129.00
100 μL
€343.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€129.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

TM8, monoclonal

species reactivity

human

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.5-1 μg/mL using HEK-293 over expressing human TIMP2 cells extract.
immunofluorescence: 2.5-5 μg/mL using human HeLa cells.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TIMP2(7077)

General description

TIMP2- Tissue Inhibitor of Metalloproteinases 2, also known as CSC-21K, is a member of the TIMP family of matrix metalloproteinases (MMPs) natural inhibitors. These proteins are involved in regulation of activated MMPs and in extracellular matrix degradation. the TIMPs family consists of 4 members which act in a non-selective manner.

Specificity

Monoclonal Anti-TIMP2 specifically recognizes human TIMP2.

Immunogen

Recombinant TIMP2 protein.

Application

The antibody is recommended to use in various immunological techniques, including Immunoblot (~24 kDa) and immunofluorescence. Detection of the TIMP2 band by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

"The MMPs activity inhibition is mediated directly by blocking the MMPs catalytic zinc ion with the TIMPs N-terminal cysteine residue.1-2 TIMP2 is also highly studied for it MMP-independent effects including cell cycle regulation 2-4 and its involvement in several cancers progression and invasiveness (e.g. renal cell carcinoma3, non-small cell lung cancer (NSCLC)5 and Gastric Cancer6).Due to the pronounced inhibitory activity, the TIMPs are considered as a potential target in designing new inhibitor variants with altered specificity for MMPs and other zinc-dependent extracellular matrix (ECM) proteins.1-2"

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service