Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200803

Sigma-Aldrich

Anti-TFE3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Class E basic helix-loop-helix protein 33, Anti-Transcription factor E3, Anti-bHLHe33

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€165.00

€165.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
€165.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€165.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~60 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

concentration

~1.0 mg/mL

technique(s)

immunoblotting: 4-8 μg/mL using human HeLa cells extract
immunofluorescence: 2.5-5 μg/mL using mouse embryo fibroblast NIH-3T3 cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFE3(7030)

General description

Transcription factor E3 (TFE3), also known as Class E basic helix-loop-helix protein 33 (bHLHe33), is a transcriptional activator that mediates the enhancer-promoter interactions. TFE3 belongs to the MiT family of helix-loop-helix leucine zipper transcription factors, it is ubiquitously expressed and can directly associate with DNA as either homodimer or heterodimer formed with two other MiT family members, TFEB or TFEC. TFE3 serves an important role in cell growth, cell proliferation, in cellular adaptation to starvation and cellular response to ER stress. Under nutrient-rich conditions TFE3 is located in the cytoplasm, under starvation conditions or treatment with ER stressors, TFE3 rapidly translocated to nuclear, there it mediates cellular adaptation to stress by simultaneously promoting lysosomal biogenesis, autophagy induction, as well as expression of critical mitochondrial and metabolic regulators. TFE3 has been shown to participate in the transcriptional regulation of the innate immune response. in addition, it was demonstrated that pathogen infections promote TFE3 nuclear translocation, thus inducing the expression of several cytokines and chemokines.

Anti-TFE3 antibody specifically recognizes TFE3 from human and mouse origin.

Immunogen

Synthetic peptide corresponding to the human TFE3

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (∼60 kDa) and Immunofluorescence. Detection of the TFE3 band by Immunoblotting is specifically inhibited by the immunizing peptide.

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Other Notes

This product is for R&D use only, not for drug, household, or other uses.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiara Di Malta et al.
EMBO molecular medicine, 15(5), e16877-e16877 (2023-03-30)
Birt-Hogg-Dubé (BHD) syndrome is an inherited familial cancer syndrome characterized by the development of cutaneous lesions, pulmonary cysts, renal tumors and cysts and caused by loss-of-function pathogenic variants in the gene encoding the tumor-suppressor protein folliculin (FLCN). FLCN acts as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service