Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200775

Sigma-Aldrich

Anti-Glycoprotein (MDR) antibody, Mouse monoclonal

clone F4, purified from hybridoma cell culture

Synonym(s):

ABCB1, ATP-binding cassette sub-family B member 1, Multidrug resistance protein 1, P-glycoprotein 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€459.00

€459.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1521

Request a Bulk Order

Select a Size

Change View
100 μL
€459.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€459.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1521

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

F4, monoclonal

form

buffered aqueous solution

species reactivity

hamster, human

concentration

~1 mg/mL

technique(s)

flow cytometry: suitable
immunoblotting: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ABCB1(5243)

General description

Anti-P-Glycoprotein (MDR) antibody, mouse monoclonal (mouse IgG1 isotype) is derived from the hybridoma F4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Multidrug resistance protein 1 (MDR1) is a member of the adenosine triphosphate (ATP)-binding cassette (ABC) transporters superfamily. It is ubiquitously expressed in kidneys, intestines, placenta, liver, adrenal glands blood-brain barrier (BBB), placenta and testis.

Specificity

Anti-P-Glycoprotein (MDR) antibody, Mouse monoclonal recognizes an epitope located at the N-terminal region of P-glycoprotein (Pgp), at the third extracellular loop of the molecule, also known as ABCB1 protein. The antibody specifically detects MDR1 P-glycoprotein, but does not recognize MDR3. Anti-P-Glycoprotein (MDR) cross reacts with MDR1 P-glycoprotein from human and hamster origin.

Immunogen

a mixture of human and hamster drug-resistant whole cells and crude plasma membranes.

Application

Anti-Glycoprotein (MDR) antibody, mouse monoclonal may be used in:
  • immunoblotting
  • immunoprecipitation
  • immunocytochemistry
  • immunohistochemistry
  • cellular enzyme-linked immunosorbent assay (ELISA)
  • flow cytometry (FACS)
  • cell surface radioimmunoassay (RIA)

Biochem/physiol Actions

Multidrug resistance protein 1 (MDR1) functions as an adenosine triphosphate (ATP)-dependent efflux pump. It is also involved in extruding certain xenobiotics and protect cells from toxicants. MDR1 gene is overexpressed in different cancer types including leukemias, lymphomas, myelomas, breast cancer, ovarian cancer, gastrointestinal stromal tumor (GIST), non-small cell lung cancer (NSCLC), fallopian tube, colon, renal, and thyroid cancers. Overexpression of MDR1 gene plays an important role in prompting multidrug resistance in cancer chemotherapy causing high resistance of cancerous cells to a wide variety of substrate anticancer drugs.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xing-Duo Dong et al.
Frontiers in pharmacology, 13, 843829-843829 (2022-03-15)
The KRAS-G12C inhibitor ARS-1620, is a novel specific covalent inhibitor of KRAS-G12C, possessing a strong targeting inhibitory effect on KRAS-G12C mutant tumors. Overexpression of ATP-binding cassette super-family B member 1 (ABCB1/P-gp) is one of the pivotal factors contributing to multidrug
Multidrug resistance in breast cancer: a meta-analysis of MDR1/gp170 expression and its possible functional significance
Trock B J, et al.
Journal of the National Cancer Institute, 89(13), 917-931 (1997)
MDR1 genotype-related pharmacokinetics and pharmacodynamics
Sakaeda T, et al.
Biological & Pharmaceutical Bulletin, 25(11), 1391-1400 (2002)
ATP hydrolysis-dependent multidrug efflux transporter: MDR1/P-glycoprotein.
Kimura Y, et al.
Current Drug Metabolism, 5(1), 1-1 (2004)
Overexpression of ABCB1 Transporter Confers Resistance to mTOR Inhibitor WYE-354 in Cancer Cells
Wang J, et al.
International Journal of Molecular Sciences, 21(4), 1387-1387 (2020)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service