Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200772

Sigma-Aldrich

Anti-BRAF (V600E) antibody produced in rabbit

enhanced validation

affinity isolated antibody

Synonym(s):

Anti-Proto-oncogene B-Raf, p94, Anti-Serine/threonine-protein kinase B-raf, Anti-v-Raf murine sarcoma viral oncogene homolog B1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€446.00

€446.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€446.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.44

€446.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~95 kDa

species reactivity

human

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using extract of human HEK-293T cells over-expressing BRAF mutant (V600E) protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BRAF(673)

General description

B-Raf proto-oncogene serine/threonine-protein kinase (BRAF), also known as Serine/threonine-protein kinase B-raf, Proto-oncogene B-Raf, p94, v-Raf murine sarcoma viral oncogene homolog B1, is a member of the RAF family. BRAF is one of the key factors in mitogen-activated protein kinase (MAPK) signaling pathway, which is activated by members of the Ras family upon growth factor-induced stimulation. BRAF controls and regulates numerous essential cellular mechanisms including cell proliferation, differentiation, development, survival, apoptosis and secretion.1-2 In contrast to the majority of oncogenic fusion kinases such as ALK, ROS1, NTRK1 and RET that are receptor tyrosine kinases, BRAF encodes a non-receptor serine/threonine kinase, placing its normal protein localization within the cytoplasm rather than associated with the plasma membrane.3
BRAF is mutated at a high frequency in several cancers, including non-Hodgkin lymphoma, colorectal cancer, malignant melanoma, thyroid carcinoma, nonsmall cell lung carcinoma, and adenocarcinoma of lung.

Specificity

Anti-BRAF (V600E) recognizes V600E mutated human BRAF protein. The antibody may be used in various immunochemical techniques including Immunoblotting.

Immunogen

Synthetic peptide from the C-terminal region of human BRAF protein, conjugated to KLH

Application

Immunoblotting: a working concentration of 2-4 g/mL is recommended using extract of human HEK-293T cells over-expressing BRAF mutant (V600E) protein.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service