Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200769

Sigma-Aldrich

Anti-Human IgG3-Peroxidase antibody, Mouse monoclonal

clone HP-6050, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin G3

Sign Into View Organizational & Contract Pricing

Select a Size

1 VIAL
€407.00

€407.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 VIAL
€407.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46

€407.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HP-6050, monoclonal

form

lyophilized powder

species reactivity

human

concentration

~2 mg/mL

technique(s)

direct ELISA: 1:80,000-1:160,000 using 2 μg/mL human IgG3 for coating.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Human IgG consist of four subclasses (1-4) that can be recognized by the antigenic difference in their heavy chains. They constitute approximately 65, 30, 5 and 4% of the total IgG, respectively. Each subclass has different biological and physiochemical properties. The IgG subclass may be preferentially produced in response to different antigens and pathological conditions. For instance, anti-polysaccharide responses are mainly of the IgG2 subclass while protein antigens responses give rise to IgG1 and IgG3 antibodies. IgG1 and IgG3 are the only subclasses capable of adherence to mononuclear phagocytes and are recognized readily by the Fc receptors on various reticulo-endothelial cells, while IgG2 and IgG4 are far less efficient. The abundance of the different IgG subclasses in the bloodstream varies with age, IgG1 and IgG3 reach normal adult levels by 5-7 years of age while IgG2 and IgG4 levels rise more slowly, reaching adult levels at about 10 years of age. Serum IgG subclass deficiencies have been recorded for different patient groups. IgG3 deficiency has been associated with a patient′s history of recurrent infectious that may lead to chronic lung disease. Decreased IgG3 levels are frequently associated with IgG1 deficiency. Examination of the distribution pattern of IgG subclasses in different types of diseases may provide insight into the related immunological processes and may assist in the diagnosis of various disorders.

Immunogen

Human IgG3 myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads

Application

Direct ELISA: a working dilution of 1:80,000-1:160,000 is recommended using 2 μg/mL human IgG3 for coating.

Physical form

Supplied as a lyophilized powder.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J C Lima-Junior et al.
Vaccine, 29(9), 1801-1811 (2011-01-11)
The Plasmodium vivax Merozoite Surface Protein-3α (PvMSP-3α) is considered as a potential vaccine candidate. However, the detailed investigations of the type of immune responses induced in naturally exposed populations are necessary. Therefore, we aim to characterize the naturally induced antibody

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service