Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200714

Sigma-Aldrich

Anti-Heat Shock Protein 70 (HSP70) antibody, Mouse monoclonal

clone BRM-22, purified from hybridoma cell culture

Synonym(s):

Anti-HSP70, Anti-HSPA1A

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€436.00

€436.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€436.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€436.00


Check Cart for Availability

Request a Bulk Order

antibody form

purified from hybridoma cell culture

Quality Level

antibody product type

primary antibodies

clone

BRM-22, monoclonal

form

buffered aqueous solution

mol wt

antigen ~70 kDa

species reactivity

plant, bovine, rabbit, chicken, human, fish, insect, monkey, dog, rat, sea urchin, mouse

concentration

~1 mg/mL

technique(s)

ELISA: suitable
dot blot: suitable
immunoblotting: 0.25-0.5 μg/mL using HeLa cell line extract
immunofluorescence: suitable
immunohistochemistry: suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HSPA1A(3303)
mouse ... HSPA1A(193740)
rat ... HSPA1A(24472)

General description

Anti-heat shock protein 70 (HSP70) antibody, mouse monoclonal (mouse IgG1 isotype) is derived from the BRM-22 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mouse immunized with purified bovine brain HSP70. Monoclonal Anti-heat shock protein 70 (HSP70) specifically recognizes HSP70 from bovine, mouse, monkey, dog, human, rat, chicken,octopus, insects, plant, fish, rabbit and sea urchin origin.
HSP70 / heat shock protein family A (Hsp70) member 1A (HSPA1A), is encoded by the gene mapped to human chromosome 6p21.33. It is widely expressed in all tissues. The encoded protein is characterized with an N-terminal ATPase domain of 45kDa and a C-terminal substrate binding domain of 25kDa which is further subdivided into a β-sandwich subdomain of 15kDa and a C-terminal α-helical subdomain.

Immunogen

purified bovine brain HSP70

Biochem/physiol Actions

The members of Hsp70 protein family are implicated in various cellular processes and are required for the response to environmental changes and survival. These proteins are essential constituents of the cellular network of molecular chaperones and folding catalysts. Hsp70 plays a vital role in several folding processes, such as the folding and assembly of newly synthesized proteins, refolding of misfolded and aggregated proteins. In addition, it also facilitates membrane translocation of organellar and secretory proteins and regulate the activity of regulatory proteins. Aberrations in the gene expression leads to skin color diversity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Honghong Zhang et al.
Oncology letters, 17(6), 5601-5607 (2019-06-13)
To the best of our knowledge, the microRNA (miR/miRNA) expression profile of plasma exosomes in ovarian cancer has not been previously studied. The aim of the present study was to investigate the practicality of using plasma exosomal miRNAs as novel
Variation in Hsp70-1A expression contributes to skin color diversity
Murase D, et al.
The Journal of Investigative Dermatology, 136, 1681-1691 (2016)
Hsp70 chaperones: Cellular functions and molecular mechanism
Mayer MP and Bukau B.
Cellular and Molecular Life Sciences, 62, 670-684 (2005)
Nghi M Nguyen et al.
Cells, 12(6) (2023-03-30)
Recently, the long-term use of sedative agents in the neonatal intensive care unit (NICU) has raised concerns about neurodevelopmental outcomes in exposed neonates. Midazolam (MDZ), a common neonatal sedative in the NICU, has been suggested to increase learning disturbances and
Dániel J Kócsó et al.
Mycotoxin research, 37(2), 141-148 (2021-03-06)
The heat shock protein (Hsp70) level was assessed after 14 days of oral gavage-exposure to fumonisin B1 (FB1: 150 µg/animal/day), deoxynivalenol (DON: 30 µg/animal/day) and zearalenone (ZEN: 150 µg/animal/day), alone or in combinations (in additive manner: FD = FB1 + DON, FZ = FB1 + ZEN, DZ = DON + ZEN and FDZ = FB1 + DON + ZEN) in the liver

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service