Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200712

Sigma-Aldrich

Anti-MAPK3 antibody, Mouse monoclonal

clone mapk3-4, purified from hybridoma cell culture

Synonym(s):

Anti-ERK1, Anti-ERT2 or Anti-p44-MAPK, Anti-p44-ERK1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.44

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

mapk3-4, monoclonal

form

buffered aqueous solution

mol wt

antigen ~43 kDa

species reactivity

human, monkey

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using whole extract of human HepG2 cells
immunoprecipitation (IP): 5-10 μg/test using total extract of A431 cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPK3(5595)

General description

Anti-MAPK3 antibody, Mouse monoclonal (mouse IgG2a isotype) is derived from the mapk3-4 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mouse. Mitogen-activated protein kinase 3 (MAPK3) is a serine/threonine protein kinase. It is an essential component of the MAP kinase signal transduction pathway.

Specificity

Monoclonal Anti-MAPK3 specifically recognizes human and monkey MAPK3. Monoclonal Anti-MAPK3 does not show cross reactivity with MAPK1 (ERK2)

Immunogen

synthetic peptide from the N-terminal region of human MAPK3, conjugated to KLH.

Application

Anti-MAPK3 antibody, Mouse monoclonal may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Mitogen-activated protein (MAP) kinase signal transduction pathway participates in the regulation of a large variety of processes, including cell adhesion, cell cycle progression, cell migration, cell survival, differentiation, metabolism, proliferation and transcription. Mitogen-activated protein kinase 3 (MAPK3) is phosphorylated on Thr-202 and Tyr-204 due to activation by upstream kinases, such as MAP2K1/MEK1 and MAP2K2/MEK2, which results in translocation into the nucleus where it, in turn, phosphorylates nuclear targets. MAPK3 is known as an important target in the diagnosis and treatment of several diseases including cancer, cardiac hypertrophy and influenza A infection.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Pearson et al.
Endocrine reviews, 22(2), 153-183 (2001-04-11)
Mitogen-activated protein (MAP) kinases comprise a family of ubiquitous proline-directed, protein-serine/threonine kinases, which participate in signal transduction pathways that control intracellular events including acute responses to hormones and major developmental changes in organisms. MAP kinases lie in protein kinase cascades.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service