Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200657

Sigma-Aldrich

Monoclonal Anti-EEF1A1 antibody produced in mouse

clone EF48, purified from hybridoma cell culture

Synonym(s):

CCS-3, CCS3, EE1A1, EEF-1, EEF1A, EF-Tu, EF1A, GRAF-1EF, HNGC:16303, LENG7, PTI1, eEF1A-1, eukaryotic translation elongation factor 1 α 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€542.00

€542.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 μL
€542.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€542.00


Estimated to ship on21 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

EF48

form

buffered aqueous solution

species reactivity

bovine, mouse, rat, human, monkey, canine

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using whole extracts of HEK-293T cells.
immunofluorescence: 0.5-1 μg/mL using SW-620 cells.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EEF1A1(1915)
mouse ... Eef1a1(13627)
rat ... Eef1a1(171361)

General description

Eukaryotic translation elongation factor 1 α 1 (EEF1A1) is a cellular protein which is part of the eukaryotic elongation factor 1 (eEF1) complex. The gene encoding it is localized on human chromosome 6q14.1.[1]
Monoclonal Anti-EEF1A1 (mouse IgG2b isotype) is derived from the hybridoma EF48 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Eukaryotic elongation factor 1 A-1 (EEF1A1), also known as EF-l α is a GTP-binding protein. EEF1A1 is one of the most abundant cytoplasmic proteins and is ubiquitously expressed except in neurons and muscle.

Immunogen

a synthetic peptide corresponding to an internal sequence of human EEF1A1

Application

Monoclonal Anti-EEF1A1 antibody produced in mouse has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Eukaryotic elongation factor 1 A-1 (EEF1A1) is involved in proper codon-anticodon binding interactions between mRNAs and tRNAs at the tRNA acceptor site of the ribosome. EEF1A also plays a translation-independent role in embryogenesis, senescence, oncogenic transformation, cell proliferation, apoptosis, cytoskeletal organization and protein degradation. Overexpression of EEF1A1 has been observed in various tumors of the pancreas, colon, breast, lung, and stomach.
Eukaryotic translation elongation factor 1 α 1 (EEF1A1) binds to aminoacetylated tRNAs and delivers them to the elongating ribosome during protein synthesis. It aids in the association of the eukaryotic elongation factor 1 (eEF1) complex with the HIV-1 (human immunodeficiency virus-1) reverse transcription complex. EEF1A1 also associates with genomic RNA secondary structures of RNA viruses and helps in their replication.[1]

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The ubiquitin-like protein FAT10 stabilizes eEF1A1 expression to promote tumor proliferation in a complex manner
Liu X, et al.
Cancer research, 76(16), 4897-4907 (2016)
A Lamberti et al.
Amino acids, 26(4), 443-448 (2004-08-04)
An increasing number of evidences suggest the involvement of the eukaryotic elongation factor 1A, a core component of the protein synthesis machinery, at the onset of cell transformation. In fact, eEF1A is shown to be up-regulated in cell death; moreover
Highly homologous eEF1A1 and eEF1A2 exhibit differential post-translational modification with significant enrichment around localized sites of sequence variation
Soares DC and Abbott CM
Biology Direct, 8(1), 29-29 (2013)
Molecular characterization and expression patterns of porcine eukaryotic elongation factor 1 A
H.L. Wang
Asian-Australasian Journal of Animal Sciences, 19(7), 953-953 (2006)
Dongsheng Li et al.
Virology journal, 12, 118-118 (2015-08-06)
The cellular protein eukaryotic translation elongation factor 1A (eEF1A) binds to aminoacylated transfer RNAs and delivers them to the ribosome during translation. eEF1A also binds to RNA secondary structures present in genomes of several RNA viruses and plays important roles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service