Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200650

Sigma-Aldrich

Monoclonal Anti-SH3GL1 (C-terminal) antibody produced in mouse

clone Ep2.17, purified from hybridoma cell culture

Synonym(s):

Anti-CNSA1, Anti-EEN, Anti-SH3D2B, Anti-SH3P8

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€488.00

€488.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€488.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€488.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Ep2.17, monoclonal

form

buffered aqueous solution

mol wt

~41 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 2-5 μg/test using using HeLa cells.
immunoblotting: 1-2 μg/mL using whole extracts of HeLa.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SH3GL1(6455)

Related Categories

General description

Monoclonal Anti-SH3GL1 (C-terminal) (mouse IgG2b isotype) is derived from the hybridoma Ep2.17 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. SH3GL1 (SH3-domain GRB2-like 1), also known as endophilin-A2, is a member of the endophilin family of Src homology 3 (SH3) domain-containing proteins. The SH3 domain drives protein-protein interactions through binding to proline-rich ligands. The SH3GL proteins have a novel N-terminal segment, and a peculiar tissue distribution: SH3GL1 is ubiquitous, SH3GL2 located in the brain, and SH3GL3 in brain and testis.

Immunogen

a synthetic peptide corresponding to a sequence at the C-terminal region of human SH3GL1

Application

Monoclonal Anti-SH3GL1 (C-terminal) antibody produced in mouse has been used in immunoblotting and flow cytometry.

Biochem/physiol Actions

SH3GL1 is involved in several brain tumors, such as glioma, where its levels were increased in proportion to the tumor progression. In cells generated from children malignant brain tumor medulloblastoma (MB), SH3GL1 expression was found to be regulated by the miRNA, miR-218.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jipeng Shi et al.
Molecular medicine reports, 8(4), 1111-1117 (2013-08-24)
An increasing number of studies have suggested that microRNAs (miRNAs) are aberrantly expressed in numerous types of tumors and that a deregulation in miRNA expression may lead to carcinogenesis. Although miR‑218 has been demonstrated to be downregulated in several types
Endophilin A2: a potential link to adiposity and beyond
Alfadda AA, et al.
Molecules and cells, 40(11), 855-855 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service