Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200644

Sigma-Aldrich

Monoclonal Anti-C10orf54 antibody produced in mouse

clone ORF.4, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
€488.00

€488.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
€488.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€488.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ORF.4, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 10 μg/test using human platelets
immunohistochemistry: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Related Categories

General description

Monoclonal Anti-C10orf54 (mouse IgG1 isotype) is derived from the hybridoma OR.4 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide.C10orf54 (Chromosome 10 open reading frame 54) is also referred to as GI24, B7 member B7-H5, VISTA (V-domain Ig suppressor of T cell activation) and Dies1. It is a novel surface glycoprotein belonging to the B7 immunoglobulin superfamily. It consists of a single IgV domain with 3 additional cysteine residues and a cytoplasmic tail. It is expressed on mature myeloid APCs (antigen-presenting cells) including peritoneal macrophages, mature BMDC (bone marrow-derived dendritic cell), neutrophils and CD11c+ DCs, and to a lesser quantity in T cells and activated Treg cells.

Immunogen

synthetic peptide corresponding to a sequence at the internal region of human C10orf54

Application

Phosphotyrosine antibody inhibitor has been used in flow-cytometry and immunohistochemistry.

Biochem/physiol Actions

Studies suggest that C10orf54 (Chromosome 10 open reading frame 54) functions as a co-inhibitory ligand and restricts the mechanism of T cell proliferation and cytokine production. It was also suggested that C10orf 54 acts as a positive regulator as well as a substrate of MT1-MMP (trans-membrane type 1-matrix metalloproteinase) during tumor growth of collagen matrix, which regulates invasive growth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Keunok Jung et al.
Immune network, 13(5), 184-193 (2013-11-08)
Co-signaling molecules are surface glycoproteins that positively or negatively regulate the T cell response to antigen. Co-signaling ligands and receptors crosstalk between the surfaces of antigen-presenting cells (APCs) and T cells, and modulate the ultimate magnitude and quality of T
Li Wang et al.
The Journal of experimental medicine, 208(3), 577-592 (2011-03-09)
The immunoglobulin (Ig) superfamily consists of many critical immune regulators, including the B7 family ligands and receptors. In this study, we identify a novel and structurally distinct Ig superfamily inhibitory ligand, whose extracellular domain bears homology to the B7 family
Moustafa A Sakr et al.
Cancer science, 101(11), 2368-2374 (2010-07-30)
GI24, an immunoglobulin superfamily member, has been cloned from a placenta cDNA library as a gene product that promoted activation of matrix metalloproteinase (MMP)-2 mediated by membrane type (MT) 1-MMP. Co-expression of GI24 with MT1-MMP in HEK293T cells increased the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service