Skip to Content
Merck
All Photos(1)

Key Documents

SAB4200590

Sigma-Aldrich

Anti-WHAMM (C-terminal) Produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€409.00

€409.00


Estimated to ship on17 May 2025


Request a Bulk Order

Select a Size

Change View
200 μL
€409.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€409.00


Estimated to ship on17 May 2025


Request a Bulk Order

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

form

buffered aqueous solution

mol wt

~100 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 1-2 μg/mL using whole extracts of HEK-293T cells overexpressing human WHAMM.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WHAMM(123720)

General description

WASP homologue associated with actin, membranes, and microtubules (WHAMM) is a nucleation promoting factor. It is located in the cis-Golgi and tubulo-vesicular membrane transport intermediates. WHAMM contains an N-terminal region, a coiled-coil region and a WCA region. It is located on the human chromosome at 15q25.2.

Specificity

Anti- WHAMM (C-terminal) recognizes human WHAMM.

Immunogen

peptide corresponding to the C-terminal region of human WHAMM , conjugated to KLH.

Application

Anti-WHAMM (C-terminal) produced in rabbit may be used in immunoblotting.

Biochem/physiol Actions

WASP homologue associated with actin, membranes, and microtubules (WHAMM) takes part in actin-related protein 2 & 3 (Arp2/3)-mediated actin polymerization, mediates Golgi membrane association and microtubule dynamics. It also takes part in the maintenance of Golgi structure and participates in vesicle transport between the endoplasmic reticulum and the Golgi apparatus.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Seminars in cell & developmental biology
Seminars in Cell & Developmental Biology, 24(4), 258-266 (2013)
Kenneth G Campellone et al.
Cell, 134(1), 148-161 (2008-07-11)
The Arp2/3 complex is an actin nucleator that plays a critical role in many cellular processes. Its activities are regulated by nucleation-promoting factors (NPFs) that function primarily during plasma membrane dynamics. Here we identify a mammalian NPF called WHAMM (WASP

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service