Skip to Content
Merck
All Photos(3)

Key Documents

SAB4200384

Sigma-Aldrich

Anti-SMAD6 (N-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-MADH6, Anti-MADH7, Anti-SMAD family member 6

Sign Into View Organizational & Contract Pricing

Select a Size

200 μL
€475.00

€475.00


Check Cart for Availability


Select a Size

Change View
200 μL
€475.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€475.00


Check Cart for Availability

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~60 kDa

species reactivity

human, mouse

concentration

~1.0 mg/mL

technique(s)

immunoprecipitation (IP): 2.5-5 μg using lysates of HEK-293T cells over-expressing human SMAD6
indirect immunofluorescence: 5-10 μg/mL using mouse 3T3 cells
western blot: 1-2 μg/mL using whole extracts of HEK-293T cells over-expressing human SMAD6

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SMAD6(4091)
rat ... Smad6(367100)

General description

SMAD6 is a member of inhibitory Smads (I-Smad) family. It consists of two conserved domains, the N-terminal MH1 and C-terminal MH2, that are separated by a proline-rich linker region of varying length.

Immunogen

peptide corresponding to the N-terminal region of human SMAD6, conjugated to KLH. The corresponding sequence is identical in rat and differs by a single amino acid in mouse.

Application

Anti-SMAD6 (N-terminal) antibody produced in rabbit has been used in:
  • immunoblotting
  • immunoprecipitation
  • immunofluorescence

Biochem/physiol Actions

SMAD6 plays an important role in transforming growth factor-β (TGF-β) pathway. It regulates cell proliferation, apoptosis and differentiation. SMAD6 is a key negative regulator of bone morphogenetic protein (BMP) signaling in the cytoplasm and in the nucleus. SMAD6 inhibits SMAD signaling in the nucleus by interacting with transcriptional repressors such as HOXC-8, class I histone deacetylases (HDACs) and transcriptional corepressor C-terminal binding protein (CtBp).

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regulation of TGF-beta signaling by Smad7
Yan X, et al.
Acta biochimica et biophysica Sinica, 41(4), 263-272 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service